Granular Activated Carbon Cartridges for "Big Blue" housing
These cartridges remove organic tastes/odors and chlorine. Special design maximizes contact time and reduces channeling or bypass that is common with GAC cartridges. Included within the cartridge is a 20-micron post-filter to reduce carbon fines††.
The 20" cartridge meets NSF Standard 42 for materials.
$49.50 - $88.50USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Filter Length (in) Micron Rating
Availability Pricing
Big Blue Filter cartridge, 10", granular carbon, 20 micron EW-01512-64 Mfr # 155153-43(GAC-BB)
COMPARE
10 20
$49.50USD / Each
Big Blue Filter cartridge, 20", granular carbon, 20 micron EW-01512-66 Mfr # 155249-43(GAC-20BB)
COMPARE
20 20
$88.50USD / Each