Great Plains Economy Fuel and Water Meters
Small, powerful, and durable
  • Battery powered 4 digit display
  • Factory calibrated to water or fuel
  • Gallon or litre measurement units
  • Batch and cumulative totals
  • Ready to install
Small, powerful, microprocessor based totalizers powered by AAA batteries have large four-digit displays and mount at the nozzle for ultimate convenience. They are suited for flow ranges from 3 to 30 GPM. Nylon® models, rated to 150 PSIG, are factory calibrated for water. Aluminum models, rated to 300 PSIG are factory calibrated for fuels. Accuracy is ±5%. All mount with 1 inch female NPT fittings.
$229.50 - $235.80USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fluid Type Max Flow Rate (LPM) Wetted Materials
Availability Pricing
Great Plains Industries Flowmeter for Fuel, Aluminum, 1" NPT(F), 10 to 100 LPM EW-32485-99 Mfr # 113255-2
COMPARE
fuel and thin fluids Aluminum
$235.80USD / Each
Great Plains Industries Flowmeter for Water, Nylon, 1" NPT(F), 10 to 100 LPM EW-32486-00 Mfr # 113255-5
COMPARE
water and thin fluids 100 Nylon
$229.50USD / Each
Great Plains Industries Flowmeter for Water, Nylon, 1" ISO Thread, 10 to 100 LPM EW-32486-01 Mfr # 113255-6
COMPARE
water and thin fluids 100 Nylon
$229.50USD / Each
Great Plains Industries Flowmeter for Fuel, Aluminum, 1" ISO Thread, 10 to 100 LPM EW-32486-02 Mfr # 113255-3
COMPARE
fuel and thin fluids 100 Aluminum
$232.50USD / Each