GT Scien TOGA® Filters
TOGA® filters are designed to enhance laboratory air quality and safeguard the health of researchers
  • Utilizes physical adsorption, chemical reaction, and neutralization for superior performance
  • No ozone or nitrogen oxide emission from side reactions compared to carbon filters
  • Fully utilizes filter passage area for effective pollutant removal
  • A Types are excellent for removing VOCs, alcohol, hydrocarbons, and odors
  • B Types offer efficient removal of general organic compounds, ozone, ammonia, and bad odors
  • C Types are great for removal of nitrogen and sulfur based oxides
The TOGA® filter signifies a paradigm shift in laboratory air quality management. In comparison to common carbon filters, the TOGA® filter's multifaceted approach—utilizing physical adsorption, chemical reaction, and neutralization—offers an array of advantages, including an extended filter replacement cycle, excellent purification of harmful gases, and minimized contamination risks.
With distinct types catering to various applications, from general organic compounds to specific chemical challenges, the TOGA® filter showcases its versatility. Rigorous performance tests conducted by reputable institutions attest to its consistently high removal efficiency, making it a reliable choice for laboratories seeking superior air quality solutions
$880.00 - $1,584.00USD / Each
41 variations of this product are available.
See All Product Options Below
41 PRODUCT OPTIONS
Compareitem specifications
Compatible With Description
Availability Pricing
GT SCIEN TOGA® Puri Intelligent Filter for 81790-34, Standard EW-81790-59 Mfr # TOGA-M01DF
COMPARE
81790-34 Toxic Gas Purifier TOGA® Puri Intelligent Filter for 81790-34, Standard
$1,408.00USD / Each
GT SCIEN TOGA® Puri Intelligent Filter for 81790-34, Type A EW-81790-60 Mfr # TOGA-M01DF-A
COMPARE
81790-34 Toxic Gas Purifier TOGA® Puri Intelligent Filter for 81790-34, Type A
$1,408.00USD / Each
GT SCIEN TOGA® Puri Intelligent Filter for 81790-34, Type B EW-81790-61 Mfr # TOGA-M01DF-B
COMPARE
81790-34 Toxic Gas Purifier TOGA® Puri Intelligent Filter for 81790-34, Type B
$1,408.00USD / Each
GT SCIEN TOGA® Puri Intelligent Filter for 81790-34, Type C EW-81790-62 Mfr # TOGA-M01DF-C
COMPARE
81790-34 Toxic Gas Purifier TOGA® Puri Intelligent Filter for 81790-34, Type C
$1,408.00USD / Each
GT SCIEN TOGA® Puri Smart Pro Filter for 81790-39 EW-81790-63 Mfr # TOGA-MAC1-AIF
COMPARE
81790-39 Toxic Gas Purifier TOGA® Puri Smart Pro Filter for 81790-39
$1,408.00USD / Each
GT SCIEN TOGA® Puri Smart Filter for 81790-35, Standard EW-81790-64 Mfr # TOGA-S02D-AIF
COMPARE
81790-35 Toxic Gas Purifier TOGA® Puri Smart Filter for 81790-35, Standard
$1,584.00USD / Each
GT SCIEN TOGA® Puri Smart Filter for 81790-35, Type A EW-81790-65 Mfr # TOGA-S02D-AIF-A
COMPARE
81790-35 Toxic Gas Purifier TOGA® Puri Smart Filter for 81790-35, Type A
$1,584.00USD / Each
GT SCIEN TOGA® Puri Smart Filter for 81790-35, Type B EW-81790-66 Mfr # TOGA-S02D-AIF-B
COMPARE
81790-35 Toxic Gas Purifier TOGA® Puri Smart Filter for 81790-35, Type B
$1,584.00USD / Each
GT SCIEN TOGA® Puri Smart Filter for 81790-35, Type C EW-81790-67 Mfr # TOGA-S02D-AIF-C
COMPARE
81790-35 Toxic Gas Purifier TOGA® Puri Smart Filter for 81790-35, Type C
$1,584.00USD / Each
GT SCIEN TOGA® Puri Cabinet Filter for 81790-54, Standard EW-81790-68 Mfr # TOGA-SCD0PF
COMPARE
81790-54 Toxic Gas Purifier TOGA® Puri Cabinet Filter for 81790-54, Standard
$1,232.00USD / Each
GT SCIEN TOGA® Puri Cabinet Filter for 81790-54, Type A EW-81790-69 Mfr # TOGA-SCD0PF-A
COMPARE
81790-54 Toxic Gas Purifier TOGA® Puri Cabinet Filter for 81790-54, Type A
$1,232.00USD / Each
GT SCIEN TOGA® Puri Cabinet Filter for 81790-54, Type B EW-81790-70 Mfr # TOGA-SCD0PF-B
COMPARE
81790-54 Toxic Gas Purifier TOGA® Puri Cabinet Filter for 81790-54, Type B
$1,232.00USD / Each
GT SCIEN TOGA® Puri Cabinet Filter for 81790-54, Type C EW-81790-71 Mfr # TOGA-SCD0PF-C
COMPARE
81790-54 Toxic Gas Purifier TOGA® Puri Cabinet Filter for 81790-54, Type C
$1,232.00USD / Each
GT SCIEN TOGA® Puri Cabinet Filter for 81790-53, Standard EW-81790-72 Mfr # TOGA-SCA0PF
COMPARE
81790-53 Toxic Gas Purifier TOGA® Puri Cabinet Filter for 81790-53, Standard
$1,232.00USD / Each
GT SCIEN TOGA® Puri Cabinet Filter for 81790-53, Type A EW-81790-73 Mfr # TOGA-SCA0PF-A
COMPARE
81790-53 Toxic Gas Purifier TOGA® Puri Cabinet Filter for 81790-53, Type A
$1,232.00USD / Each
GT SCIEN TOGA® Puri Cabinet Filter for 81790-53, Type B EW-81790-74 Mfr # TOGA-SCA0PF-B
COMPARE
81790-53 Toxic Gas Purifier TOGA® Puri Cabinet Filter for 81790-53, Type B
$1,232.00USD / Each
GT SCIEN TOGA® Puri Cabinet Filter for 81790-53, Type C EW-81790-75 Mfr # TOGA-SCA0PF-C
COMPARE
81790-53 Toxic Gas Purifier TOGA® Puri Cabinet Filter for 81790-53, Type C
$1,232.00USD / Each
GT SCIEN TOGA® Puri Smart Filter for 81790-40, Standard EW-81790-76 Mfr # TOGA-M01AIF
COMPARE
81790-40 Toxic Gas Purifier TOGA® Puri Smart Filter for 81790-40, Standard
$1,408.00USD / Each
GT SCIEN TOGA® Puri Smart Filter for 81790-40, Type A EW-81790-77 Mfr # TOGA-M01AIF-A
COMPARE
81790-40 Toxic Gas Purifier TOGA® Puri Smart Filter for 81790-40, Type A
$1,408.00USD / Each
GT SCIEN TOGA® Puri Smart Filter for 81790-40, Type B EW-81790-78 Mfr # TOGA-M01AIF-B
COMPARE
81790-40 Toxic Gas Purifier TOGA® Puri Smart Filter for 81790-40, Type B
$1,408.00USD / Each
Showing 1 - 20 of 41