GT Scien TOGA® Fridge Smart Chemical Storage Refrigerators
Introducing the TOGA® Fridge Smart refrigerator, a completely enclosed solution for chemical storage, seamlessly integrating refrigeration and toxic gas removal for enhanced safety and efficiency in laboratories
  • Dual intake blowers and an internal circulation blower ensure uniform temperature across all storage levels
  • Patented TOGA® filter removes harmful gases from the reagent chamber
  • AIoT technology facilitates real-time management of chemicals, equipment monitoring, and automated operation of networked devices
  • Dual layer tempered and heated glass enhances insulation and prevents condensation, ensuring temperature reliability
  • Castors with adjustable stoppers provide mobility and easy leveling of equipment
  • Sliding shelf enables safe and easy handling of chemicals
  • 7" LCD Touch screen for real-time display, internal temperature control, and monitoring of chemical information
The TOGA® Fridge Smart chemical storage refrigerators are a novel solution for labs needing to store refrigerated chemicals. The innovative refrigeration design ensures a uniform temperature across all storage levels, complemented by the TOGA® filter to remove harmful gases. The integration of AIoT technology takes safety and efficiency to the next level, allowing for real-time management of chemicals. AIoT key features offer a robust set of tools for user access management, chemical tracking, and more. Users can review stored chemical information, get notified of chemical expiration, classify chemicals by hazard class, and upload SDS’s. The portal can be accessed via web or app.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Number of Doors Power (VAC) Refrigerator Capacity (Liters)
Availability Pricing
GT SCIEN TOGA® Fridge Smart Single Door Chemical Storage Refrigerator; 220V EW-81790-57 Mfr # TOGA-UGSR11P
COMPARE
1 220 500
Call For Price
GT SCIEN TOGA® Fridge Smart Double Door Chemical Storage Refrigerator; 220V EW-81790-58 Mfr # TOGA-UGSR12P
COMPARE
2 220 1050