Guzzler® Diaphragm Hand Pump Hoses
Optimize performance of your Guzzler® Diaphragm Hand Pump
These hoses are noncollapsing, wire-reinforced, and constructed of durable vinyl.
Model 06309-10 hose is compatible with Guzzler pumps 07090-09 and -10.
Model 06309-11 hose is compatible with Guzzler pump 07090-11.
Model 06309-12 hose is compatible with Guzzler pump 07090-12.
$48.75 - $141.50USD / Pkg of 1
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Tube Length, Inlet (in)
Availability Pricing
Guzzler 44 Hose for model 07090-10 (Tube length: 192)
Always in Stock
EW-06309-10 Mfr # SG.H.-1
COMPARE
192
$48.75USD / Pkg of 1
Guzzler 45 Hose for model 07090-11 (Tube length: 192) EW-06309-11 Mfr # SG.H.-1 1/2
COMPARE
192
$65.50USD / Pkg of 1
Guzzler 46 Corregated Vinyl Hose, 2" ID; 12 ft Length EW-06309-12 Mfr # SG.H.-2
COMPARE
192
$141.50USD / Pkg of 1