This product is now discontinued, View Related Products
GW Instek AFG-2000 Single- and Dual-Channel Arbitrary Function Generators
Greatly expand your application ranges
  • Simultaneously displays amplitude, DC offset, and more on the 3.5” LCD
  • Adjustable duty cycle allows you to use units as a pulse generator to create pulse waveforms
  • Built-in frequency counter eliminates the need to purchase a separate frequency counter
  • Free downloadable software provides a powerful analysis tool
Economical arbitrary function generators are an accurate signal source, designed to accommodate a wide range of applications typically for educational and basic industrial requirements. Up to 120 MSa/s sampling rate, 10-bit vertical resolution, and 4 k point waveform memory allow you to create the needed waveforms, point by point, through the intuitive user interface. Units feature a USB device interface for remote control and waveform editing through a computer. The free downloadable software lets you easily edit your complicated and irregular waveforms.
Call For Price
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
GW Instek AFG-2005 Function Generator, 1 Ch., 5 MHz EW-20050-55 Mfr # AFG-2005
COMPARE
Function Generator, 1 Ch., 5 MHz
Call For Price
GW Instek AFG-2012 Function Generator, 1 Ch., 12 MHz EW-20050-56 Mfr # AFG-2012
COMPARE
Function Generator, 1 Ch., 12 MHz
Call For Price
GW Instek AFG-2025 Function Generator, 1 Ch., 25 MHz EW-20050-57 Mfr # AFG-2025
COMPARE
Function Generator, 1 Ch., 25 MHz
Call For Price
GW Instek AFG-2105 Function Generator, 1 Ch., 5 MHz EW-20050-58 Mfr # AFG-2105
COMPARE
Function Generator, 1 Ch., 5 MHz
Call For Price
GW Instek AFG-2112 Function Generator, 1 Ch., 12 MHz EW-20050-59 Mfr # AFG-2112
COMPARE
Function Generator, 1 Ch., 12 MHz
Call For Price
GW Instek AFG-2125 Function Generator, 1 Ch., 25 MHz EW-20050-60 Mfr # AFG-2125
COMPARE
Function Generator, 1 Ch., 25 MHz
Call For Price
GW Instek AFG-2225 Function Generator, 2 Ch., 25 MHz EW-20050-61 Mfr # AFG-2225
COMPARE
Function Generator, 2 Ch., 25 MHz
Call For Price
Showing 1- 7 of 7