This product is now discontinued, View Related Products
GW Instek PSM Series Dual-Range Programmable DC Power Supplies
Programmable with high-precision output
  • Dual-range output up to 200 W for additional flexibility
  • High resolution and low noise provide excellent regulation
  • Overvoltage, overcurrent, and overtemperature protection for added safety
  • Up to 100 sets of save and recall memory
  • Auto step running with time setting for continuous testing
  • Self-test and software calibration reduce maintenance
  • RS-232 and GPIB standard interfaces
The PSM-series power supplies provide high-resolution, high-stability, DC programmable power with dual power ranges, offering more flexibility for power output applications. These units are ideal for university/research labs, ATE for production testing, QA verification, and product development applications.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
GW Instek PSM-2010 Dual Range DC Power Supply, 8 V/20 A, 20 V/10 A EW-20050-40 Mfr # PSM-2010
COMPARE
Dual Range DC Power Supply, 8 V/20 A, 20 V/10 A
Call For Price
GW Instek PSM-3004 Dual Range DC Power Supply, 15 V/7 A, 30 V/4 A EW-20050-41 Mfr # PSM-3004
COMPARE
Dual Range DC Power Supply, 15 V/7 A, 30 V/4 A
Call For Price
GW Instek PSM-6003 Dual Range DC Power Supply, 30 V/6 A, 60 V/3.3 A EW-20050-42 Mfr # PSM-6003
COMPARE
Dual Range DC Power Supply, 30 V/6 A, 60 V/3.3 A