Hach COD Reactors
Ideal for wastewater influent and effluent testing, industrial process water testing, and environmental monitoring
  • Uses EPA-approved COD method—requires less equipment, lab space, and maintenance than other open reflux COD tests
  • High flexibility, customization, and temperature accuracy
  • For a complete system order COD reactor (99574-60 or -65), COD or TOC test kits (99574-25,-26,-27,-28, or -29), and colorimeter (99573-00)
Hach DRB-200 COD Reactor digests up to 15 samples simultaneously. Reactors feature one-touch operation through the use of preprogrammed Hach COD and TNT standard digestion temperatures/times, or customize up to three programs. To customize, choose from one of nine languages, set the temperature between 37 to 165°C, and enter the time between 1‑minute to 8 hours. Built-in timer features audible alarm and automatic shut-off to prevent overheating. Transparent, locking lid covers 16 x 100-mm vials while protecting the user in the unlikely event of a broken reagent vial.
For full specifications and operating instructions view the instruction manual linked below.
$1,562.00 - $1,563.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
Hach LTV082.53.40001 DRB 200 COD Reactor, single block, 15 X 16 mm vial wells, 115 VAC EW-99574-60 Mfr # LTV082.53.40001
COMPARE
115
$1,562.00USD / Each
Hach LTV082.52.40001 DRB 200 COD Reactor, single block, 15 X 16 mm vial wells, 230 VAC EW-99574-65 Mfr # LTV082.52.40001
COMPARE
220
$1,563.00USD / Each