Hach HQ1110 Series Portable pH/ORP/mV Meters
A robust and intuitive pH, ORP, and mV meter
  • IntelliCal™ smart probes have “plug-and-play” automatic parameter recognition
  • Easy step-by-step illustrated calibration and troubleshooting procedures
  • Store up to 5,000 data points
  • Rugged, IP67-rated waterproof housing
  • Option to select from 27 languages in set up mode
Hach HQ portable meters offer a versatile range of measurements, including pH, ORP, mV, and temperature. The Intellical™ probes are automatically recognized by the HQ meter and will retain calibration history and method settings to minimize errors and setup time. The proven technology delivers superior accuracy and response time, even when measuring challenging samples or in the field.
The illustrated screen displays step-by-step calibration and troubleshooting procedures on the 536 x 335 Mono-TFT display. Whether you are in the lab or in the field, transfer your measurements in just a few clicks via USB or optional Bluetooth. The optional Hach Communication Dongle (HCD) connects the meter to Claros™ data management software for additional data validation.
$855.00 - $2,256.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Hach HQ1110 Portable Dedicated pH/ORP/mV Meter EW-59005-00 Mfr # LEV015.53.1110A
COMPARE
Portable Dedicated pH/ORP/mV Meter
$855.00USD / Each
Hach HQ1110 Portable Dedicated pH/ORP/mV Meter with Gel ORP Electrode, 5 m Rugged Cable EW-59005-01 Mfr # LEV015.53.11104
COMPARE
Portable Dedicated pH/ORP/mV Meter with Gel ORP Electrode, 5 m Rugged Cable
$2,256.00USD / Each
Hach HQ1110 Portable Dedicated pH/ORP/mV Meter with Low-Maintenance Gel pH Electrode, 1 m Cable EW-59005-02 Mfr # LEV015.53.11101
COMPARE
Portable Dedicated pH/ORP/mV Meter with Low Maintenance Gel pH Electrode, 1 m Cable
$1,636.00USD / Each
Hach HQ1110 Portable Dedicated pH/ORP/mV Meter with General-Purpose Gel pH Electrode, 1 m Cable EW-59005-03 Mfr # LEV015.53.11102
COMPARE
Portable Dedicated pH/ORP/mV Meter with General Purpose Gel pH Electrode, 1 m Cable
$1,691.00USD / Each
Hach HQ1110 Portable Dedicated pH/ORP/mV Meter with General-Purpose Gel pH Electrode, 5 m Rugged Cable EW-59005-04 Mfr # LEV015.53.11103
COMPARE
Portable Dedicated pH/ORP/mV Meter with General Purpose Gel pH Electrode, 5 m Rugged Cable
$2,249.00USD / Each