This product is now discontinued, View Related Products
Hach HQ40d Dual-Input Multiparameter Portable Meter Kits
pH/conductivity/oxygen meters with the convenience and accuracy of LDO luninescent technology
  • IntelliCal™ smart probes have ''plug-and-play'' automatic parameter recognition
  • Option to select from 13 languages in setup mode
  • Store up to 500 data sets with GLP-compliance
  • Rugged, IP67-rated waterproof housing
Versatile HQd meters offer a large backlit display with an intuitive user-interface. The meter prompts you through calibration, setup, and measurements. This meter has dual input and multiparameter operation for pH, conductivity, and LDO with temperature.
Unit can automatically or manually log up to 500 measurements with probe serial number, current calibration data, user ID, sample ID, time, and date. Use the USB port to download data to a PC or flash drive.
Precalibrated Hach IntelliCAL smart probes store their own calibration data and identification. The meter recognizes each probe, so you can easily move between meters without recalibrating. Choose between a meter kit with laboratory probes (59004-18) or rugged probes (59004-48) depending on your application.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max Conductivity Max pH (pH)
Availability Pricing
Hach 8506000 Hq40D Portable PH/Con/LDO Laboratory Meter Kit EW-59004-18 Mfr # 8506000
COMPARE
14
Call For Price
Hach 8507400 ORP/DO FIELD KIT EW-59004-44 Mfr # 8507400
COMPARE
Call For Price
Hach 8505300 Hqd Portable PH/Con/LDO Field Meter Kit EW-59004-48 Mfr # 8505300
COMPARE
14