This product is now discontinued, View Related Products
Hach sensION + Waterproof pH/mV Meter
Basic all-in-one kits with everything you need to start testing
  • Guided menu navigation makes pH measurements fast and simple
  • Automatic buffer recognition at 2.00, 4.01, 7.00, 9.21 and 10.01
  • IP67-rated waterproof housing
The sensIon+ pH, mV and temperature meter is packaged with everything you need to start testing clean water or wastewater. Meters feature a backlit graphic LCD with easy to follow Icon-based user interface.
Other features include selectable auto-off, two point temperature re-adjustment, ORP calibration, a time measurement indicator for electrode life, and improved stabilization algorithm. The hard carrying case also acts as a workstation in the field.
Clean/Drinking water PH1 kit includes general pH/ATC electrode 59004-53.
Wastewater PH1 kit includes double junction pH/ATC electrode 59004-54.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Max pH (pH) Description
Availability Pricing
Hach PH1 5050T SensION+ Drinking Water pH Meter Kit EW-59004-01 Mfr # LPV2550T.97.002
COMPARE
14 SensION+ Drinking Water pH Meter Kit
Call For Price
Hach PH1 5051T SensION+ Wastewater pH Meter Kit EW-59004-03 Mfr # LPV2551T.97.002
COMPARE
14 SensION+ Wastewater pH Meter Kit
Call For Price
Hach METER PH/MV/ T EW-59004-04 Mfr # LPV2010T.97.002
COMPARE
Call For Price
Hach METER PH/ISE/ MV/ T EW-59004-06 Mfr # LPV2210T.97.002