Hand-Operated Piston Dispensing Drum Pump, PVC
Economical piston pumps are ideal for a variety of fluids
  • Adjustable to most pails and drums
Models 07079-50 and -55 feature an angled spout to help prevent dripping. Models 07079-60 and -65 are made entirely of PVC, including the piston and seal, making them an ideal option for sanitary applications.
$62.45 - $127.20USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Container Adapter Flow Rate (Strokes/Gal) Outlet Connection
Availability Pricing
COMPARE
3/4" and 2" IPS Bung 16 Spout
$62.45USD / Each
COMPARE
3/4" and 2" IPS Bung 6 Spout
$118.20USD / Each
COMPARE
2" IPS Bung 16 Hose
$66.90USD / Each
COMPARE
2" IPS Bung 6 Hose
$127.20USD / Each