Hand-Operated Siphon Drum Pump Accessories
Compatible with Siphon and Piston Hand-Operated Pumps
  • Choose from a wide range of replacement parts and accessories
Model 07079-10 steel screw in adapter features a 1-3/4" NPT(F) connection and fits 5 gallon pails.
Model 07079-15 flex spout adapter is constructed of durable polyethylene and fits 5 gallon containers with 5-5/8" (6.7 cm) openings.
Model 07079-17 bung converter kit converts a Mauser course bung thread or standard course buttress to a 2" NPT connection.
$20.50 - $25.50USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Steel screw—in adapter, 1-3/4" NPT(F)
$24.25USD / Each
COMPARE
Flex Spout Adapter for 5-Gallon Containers with 2-5/8" Opening, polyethylene
$20.50USD / Each
COMPARE
Bung converter kit converts Mauser or standard course buttress to 2" NPT
$25.50USD / Each