COMARK Handheld Differential Pressure Gauges
Intrinsically safe models are ideal for refineries, petrochemical plants, and mines - certified EEx ia IIC T4
  • Select one of nine units
  • Features zero, hold, max., min., out of range, and low battery functions
  • Smooth fluctuating readings with filter functions
  • Displays average of sixteen pressure readings
What's included: one 9 V battery, two 2-ft lengths of nylon tubing. Intrinsically safe models also include an allen wrench (for battery replacement), and a hard plastic carrying case.
ITEM(S) ON SALE
$609.42 - $708.41USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Resolution Description
Availability Pricing
COMARK Handheld Differential Pressure Gauge, 0 to 2100 mbar, Intrinsically Safe EW-68600-02 Mfr # C9505/IS
COMPARE
0.1 mbar Handheld Differential Pressure Gauge, 0 to 2100 mbar, Intrinsically Safe
$708.41USD / Each
$885.52USD / Each
COMARK Handheld Differential Pressure Gauge, 0 to 7000 mbar, Intrinsically Safe EW-68600-04 Mfr # C9507/IS
COMPARE
1 mbar Handheld Differential Pressure Gauge, 0 to 7000 mbar, Intrinsically Safe
$707.64USD / Each
$884.56USD / Each
COMARK Handheld Differential Pressure Gauge, 0 to 350 mbar, Intrinsically Safe EW-68600-06 Mfr # C9503/IS
COMPARE
0.1 mbar Handheld Differential Pressure Gauge, 0 to 350 mbar, Intrinsically Safe
$697.04USD / Each
$871.30USD / Each
COMARK Handheld Differential Pressure Gauge, 0 to 350 mbar, Dry Use EW-68600-10 Mfr # C9553
COMPARE
0.1 mbar Handheld Differential Pressure Gauge, 0 to 350 mbar, Dry Use
$609.42USD / Each
$761.78USD / Each
COMARK Handheld Differential Pressure Gauge, 0 to 2100 mbar, Dry Use EW-68600-12 Mfr # C9555
COMPARE
0.1 mbar Handheld Differential Pressure Gauge, 0 to 2100 mbar, Dry Use
$609.42USD / Each
$761.78USD / Each
COMARK Handheld Differential Pressure Gauge, 0 to 7000 mbar, Dry Use EW-68600-14 Mfr # C9557
COMPARE
1 mbar Handheld Differential Pressure Gauge, 0 to 7000 mbar, Dry Use
$609.42USD / Each
$761.78USD / Each