Harwil In-Line Liquid Level Switches
Eliminate nuisance switch tripping with a wide range of differentials
  • Large differentials are immune to wave action and turbulence (L-21VCR models)
  • Single-point switch provides pump up/pump down level control
  • Withstands corrosive and particle-contaminated working fluids
This series of liquid level switches are easy to install, operate and maintain. Through-wall NPT process fittings and NPT electrical ports facilitate integration into existing systems. Operation checks and troubleshooting require only a multimeter.
Wetted materials are fiberglass-reinforced PPS and Hastelloy® C. Switch types and materials of construction resist corrosion and increase durability, making them suitable even for seawater and filtered sewage.
$107.99 - $274.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Min Secific Gravity
Availability Pricing
Harwil In-Line Liquid Level Switch, 250 psi, 1-1/4" NPT, 1" Differential EW-34001-55 Mfr # L-21VCR/15A/1/F
COMPARE
250 0.7
$225.20USD / Each
Harwil In-Line Liquid Level Switch, 250 psi, 1-1/4" NPT, 2" Differential EW-34001-57 Mfr # L-21VCR/15A/2/F
COMPARE
250 0.7
$227.20USD / Each
Harwil In-Line Liquid Level Switch, 250 psi, 1-1/4" NPT, 3" Differential EW-34001-59 Mfr # L-21VCR/15A/3/F
COMPARE
250 0.7
$227.20USD / Each
Harwil In-Line Liquid Level Switch, 250 psi, 1-1/4" NPT, 5" Differential EW-34001-61 Mfr # L-21VCR/15A/5/F
COMPARE
250 0.7
$225.20USD / Each
Harwil L-30CR/F In-Line Liquid Level Switch, 1/2" NPT(M) EW-34001-63 Mfr # L-30CR/F
COMPARE
75 0.8
$274.00USD / Each
Harwil Liquid Level Switch, normally open (N.O.), horizontal mount EW-34001-65 Mfr # L-40VCR/NO/HOR
COMPARE
200 0.7
$107.99USD / Each