Hayward Electrically Actuated Reversing Ball Valves
Quarter-turn valves offer numerous standard features
  • Four limit switches provide electrical control of actuator
  • NEMA 4X electrical enclosure
  • Position indicators show yellow for open and red for closed valve
  • Clutchless manual override allows you to close valve in case of power loss
  • Thermal overload protection provides auto shutdown in case of power surges
$898.00 - $958.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Inlet Port Outlet Port Port Size (NPT(F))
Availability Pricing
Hayward HCTB1075STACTV/EPM2 Electrically Actuated PVC Ball Valve, 3/4" NPT(F), 120 VAC EW-07390-61 Mfr # HCTB1075STACTV/EPM2
COMPARE
3/4" true union socket 3/4" true union socket
$898.00USD / Each
Hayward HCTB1150STACTV/EPM2 Electrically Actuated PVC Ball Valve, 1 1/2" NPT(F), 120 VAC EW-07390-65 Mfr # HCTB1150STACTV/EPM2
COMPARE
1-1/2" true union socket 1-1/2" true union socket
$958.00USD / Each