Hayward NVA Series Plastic Needle Valves
  • Accurate flow control lets you adjust flow down to drops per minute
  • Seals are heavy-duty FPM O-ring
  • Natural PVDF valves are the ideal option for high-purity, high-temperature applications
Precision needle valves are available in four materials: PVC, CPVC, PP and PVDF. All feature an integrated stem with PTFE seat; fine pitch stem threads ensure precise adjustment. Valve body also includes flanges for panel mounting. Connections are standard pipe thread.
$105.00 - $177.00USD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connection Type Cv Housing Material
Availability Pricing
Hayward 155.522.554 PVC Needle Valve, 1/4" NPT(F), with PTFE and Viton EW-03245-60 Mfr # NVA1025T
COMPARE
1/4" NPT(F) 0.64 PVC
$105.00USD / Each
Hayward 155.522.554 PVC Needle Valve, 3/8" NPT(F), with PTFE and Viton EW-03245-62 Mfr # NVA1037T
COMPARE
3/8" NPT(F) 0.72 PVC
$117.00USD / Each
Hayward 155.522.554 PVC Needle Valve, 1/2" NPT(F), with PTFE and Viton EW-03245-64 Mfr # NVA1050T
COMPARE
1/2" NPT(F) 0.79 PVC
$120.00USD / Each
Hayward NV40025T PP Needle Valve, 1/4" NPT(F), with PTFE and Viton EW-03245-66 Mfr # NVA4025T
COMPARE
1/4" NPT(F) 0.64 PP
$129.00USD / Each
Hayward NV40025T PP Needle Valve, 3/8" NPT(F), with PTFE and Viton EW-03245-68 Mfr # NVA4037T
COMPARE
3/8" NPT(F) 0.72 PP
$145.00USD / Each
Hayward NV40025T PP Needle Valve, 1/2" NPT(F), with PTFE and Viton EW-03245-70 Mfr # NVA4050T
COMPARE
1/2" NPT(F) 0.79 PP
$145.00USD / Each
Hayward CPVC Needle Valve, 1/4" NPT(F), with PTFE Seat and FPM Seals EW-03245-72 Mfr # NVA2025T
COMPARE
1/4" NPT(F) 0.64 CPVC
$110.00USD / Each
Hayward CPVC Needle Valve, 3/8" NPT(F), with PTFE Seat and FPM Seals EW-03245-74 Mfr # NVA2037T
COMPARE
3/8" NPT(F) 0.72 CPVC
$125.00USD / Each
Hayward CPVC Needle Valve, 1/2" NPT(F), with PTFE Seat and FPM Seals EW-03245-76 Mfr # NVA2050T
COMPARE
1/2" NPT(F) 0.79 CPVC
$129.00USD / Each
Hayward PVDF Needle Valve, 1/4" NPT(F), with PTFE Seat and FPM Seals EW-03245-86 Mfr # NVA5025T
COMPARE
1/4" NPT(F) 0.64 PVDF
$177.00USD / Each
Hayward PVDF Needle Valve, 3/8" NPT(F), with PTFE Seat and FPM Seals EW-03245-88 Mfr # NVA5037T
COMPARE
3/8" NPT(F) 0.72 PVDF
$177.00USD / Each
Hayward PVDF Needle Valve, 1/2" NPT(F), with PTFE Seat and FPM Seals EW-03245-90 Mfr # NVA5050T
COMPARE
1/2" NPT(F) 0.79 PVDF
$177.00USD / Each
Showing 1- 12 of 12
Need Help with this product?
Call Us At1-847-549-7600
Live chat