Hayward® True Union Ball Check Valves with Socket and NPT(F) Threaded Connectors
Unique square-cut elastomer seat provides seal at very low back pressures
  • Constructed of a PVC body and EPDM seal
Quickly and easily install either vertically or horizontally. These valves are assembled with water soluble, silicone-free lubricant and constructed of a one-piece PVC body.
$57.90 - $215.80USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cv Port Size (NPT(F))
Availability Pricing
Hayward True Union Ball Check Valve, 1/2" socket and NPT(F) connectors EW-01341-00 Mfr # TC10050STE
COMPARE
4.6 0.5
$69.25USD / Each
Hayward True Union Ball Check Valve, 3/4" socket and NPT(F) connectors EW-01341-02 Mfr # TC10075STE
COMPARE
10 0.75
$81.75USD / Each
Hayward TC10100STE True Union Ball Check Valve, 1" socket and NPT(F) connectors EW-01341-04 Mfr # TC10100STE
COMPARE
28 1
$104.70USD / Each
Hayward True Union Ball Check Valve, 1-1/2" socket and NPT(F) connectors EW-01341-08 Mfr # TC10150STE
COMPARE
55 1.5
$163.50USD / Each
Hayward TC10200STE True Union Ball Check Valve, 2" socket and NPT(F) connectors EW-01341-10 Mfr # TC10200STE
COMPARE
90 2
$215.80USD / Each
GF Signet 161.360.581 Ball Check Valve, PVC, 3/8" NPT(F); 1/Each EW-01368-10 Mfr # 161561101
COMPARE
4.8 0.375
$57.90USD / Each