Hazardous-Duty Gear-Drive Air Motor Mixers
Safely use these air-drive mixers whenever electrical power is hazardous or impractical. Ideal for mixing solvents, lacquers, and volatile chemicals without the possibility of spark ignition.
Explosion proof mixers are a no-brainer when working in hazardous environments. Distilleries, refineries, and many other plants often seek explosion proof mixers for their applications. The hazards they face are real, both physical and chemical in many cases. Mixers are also a common sight in these liquid processing facilities. Putting these together, these environments require mixers with hazardous duty ratings to prove they will not create an explosion hazard. Adding in the viscosity concerns and batch sizes common to most industrial processes, a high-torque gear-drive hazardous-duty gear-drive mixer is the most common and preferred solution to mixing needs. Choosing explosion proof mixers for industrial applications is one of the most difficult engineering challenges. It is part science and part art. Consideration must be made for: viscosity, specific gravity, temperature, speed, timing, material compatibility, particulates, tank type, size, and dimensions, mission, and compliance. Our experience, training, and relationships enable us to solve your industrial mixer challenge from the R&D bench to the production floor. If you have questions about explosion proof mixers contact an Application Specialist today.
$785.00 - $1,082.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Max Torque (in-oz) Motor hp
Availability Pricing
Low-cost, gear-drive air motor mixer; 8-170 rpm EW-04681-00 Mfr # MODEL GG
COMPARE
170 1600 1/3
$1,082.00USD / Each
Low-cost, gear-drive air motor mixer with steel gears; 50-1200 rpm EW-04681-10 Mfr # G-HD
COMPARE
1200 288 1/3
$953.00USD / Each
Low-cost, gear-drive air motor mixer; 50-1200 rpm EW-04685-00 Mfr # TYPE G
COMPARE
1200 288 1/3
$785.00USD / Each
Low-cost, gear-drive air motor mixer; 50-1000 rpm EW-04686-10 Mfr # TYPE G-5
COMPARE
1000 416 1/2
$1,052.00USD / Each