HD Electric Digital Voltmeters - Phasers
Get safe and accurate voltage readings
  • Feature a backlit display for low-light conditions, auto shut-off, and low-battery indicator
  • Powered by 9 V batteries
  • True RMS voltage measurements
The DigiVolt® series of digital voltmeters/phasing sets are designed to take voltage measurements and determine if two conductors are in-phase or out-of-phase on system voltages.
$2,487.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
DC Max (Volts) DC Voltage Accuracy True Rms
Availability Pricing
HD Electric DVM-80UVM Dual Stick Digital Voltmeter, True RMS, 0-40 kV EW-23022-49 Mfr # DVM-80UVM
COMPARE
40000
$2,487.00USD / Each