This product is now discontinued, View Related Products
HD Electric High Voltage Halo Ammeter
Ideally suited for taking spot-load studies at transformers or at any mid-span location, as well as phase balance studies on primary or secondary lines. This is a true RMS reading instrument. Its load-latching circuitry continually latches and locks on to peak system currents. It will display and hold the reading until the display is reset.
Call For Price
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
DC Max (Volts) DC Voltage Accuracy True Rms
Availability Pricing
HD Electric 8280 Digital High-Voltage Ammeter EW-20021-97 Mfr # 8280
COMPARE
Call For Price
HD Electric HC-5 Aluminum Hard Case for Halo Ammeter EW-20021-98 Mfr # HC-5
COMPARE
Call For Price
HD Electric 8281 Ammeter, 0 to 2000 Amps AC, True RMS, Peak Hold, Fork Head EW-25751-01 Mfr # 8281
COMPARE
Call For Price
HD Electric 9390 Digital High-Voltage TRMS Hook Head Ammeter EW-25751-02 Mfr # 9390
COMPARE
Call For Price
HD Electric 9391 Digital High-Voltage TRMS Fork Head Ammeter EW-25751-03 Mfr # 9391