This product is now discontinued, View Related Products
HD Electric Proximity Voltage Detector
Be safe—safety tool alerts when close to dangerous high voltages
  • Dual-range unit detects voltage of 50 V to 4 kV, and 4 kV and above
  • Rugged water-resistant housing is made for outdoor use—perfect for police, fire fighters, paramedics, telecom and cable workers, and tree trimmers
  • Heartbeat-type signal ensures you that the unit is on and readily detecting voltage
  • Long-lasting rechargeable battery is included with each voltage detector
  • Dual hotstick connection accepts universal spine or shotgun types (hotstick is required; order separately)
The PRX-4 is a noncontact proximity voltage detector that audibly and visually alerts when you are located 3" (76 mm) from a 50 V source, or 10" (254 mm) from a 4 kV source or higher. The audible alarm is loud and LED lights let you know which range has been detected. Quickly recharge the battery (included) for continuous protection—a 15-minute charge lasts all day and a full (3-hour) charge lasts all week!
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
DC Max (Volts) DC Voltage Accuracy True Rms
Availability Pricing
HD Electric PRX-4 Complete Kit Proximity Voltage Detector; Car/Wall Charger, Holster, Clip, Case EW-20025-80 Mfr # PRX-4
COMPARE
Call For Price
HD Electric PRX-4 Kit Vesion 1 Proximity Voltage Detector; Car Charger EW-20025-81 Mfr # PRX-4/K01
COMPARE
Call For Price
HD Electric PRX-4 Kit Vesion 2 Proximity Voltage Detector; Car/Wall Charger EW-20025-82 Mfr # PRX-4/K02
COMPARE
Call For Price
HD Electric PRX-4 Kit Vesion 3 Proximity Voltage Detector; Car/Wall Charger, Bag EW-20025-83 Mfr # PRX-4/K03