Heat-O-Matic Immersion Heaters with Thermostat

Heaters with Thermostatic Temperature Controller have a sensitivity of ±2°>F(±1°>C) in an agitated bath. Heaters feature a coiled heating element to concentrate heat at bottom of vessel. A 6-ft, three-wire cord with plug is included. Voltage transformer listed below is required for 230 VAC operation.
$516.50 - $539.00USD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Immersion Depth (in) Max Temperature (° F) Power (VAC)
Availability Pricing
George Ulanet 492-1 Heat-O-Matic Immersion Heater with Thermostatic Controller, 1000 watts, 7" dia, 115 VAC EW-03052-10 Mfr # M492-01-00
COMPARE
5 250 115
$536.50USD / Each
George Ulanet 492-2 Heat-O-Matic Immersion Heater with Thermostatic Controller, 1500 watts, 7" dia EW-03052-20 Mfr # MS492-02-00
COMPARE
5 250 115
$533.00USD / Each
George Ulanet 492-3 Immersion Heater with Thermostatic Controller, 1000 Watts, 10-1/2" Dia, 115 V EW-03052-30 Mfr # 492-3
COMPARE
5 250 115
$516.50USD / Each
George Ulanet 492-4 Heat-O-Matic Immersion Heater with Thermostatic Controller, 1500 watts, 14" dia EW-03052-40 Mfr # MS492-04-00
COMPARE
3 250 115
$539.00USD / Each
George Ulanet 492-6 Heat-O-Matic Immersion Heater with Thermostatic Controller, 1500 watts, 13" dia, 115 VAC EW-03052-50 Mfr # 492-6
COMPARE
5 250 115
$528.00USD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat