George Ulanet Heat-O-Matic Immersion Heaters
Heaters with dial temperature controller
  • Plastic handle offers secure grip
  • Pilot light indicates when heater is on
Heaters with Dial Temperature Controller have a sensitivity of ±1°>F when liquid is stirred. Top-mounted pilot light indicates that heater is on. Choose from four styles "A" loop, "B" short rod, "C" flat, and "D" long rod which is the same as style "B" only longer. Heaters feature a plastic grip handle; and a 6-ft, three-wire cord, 115 VAC models have U.S. standard plug, 230 VAC models do not have a plug.
$264.50 - $365.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Immersion Depth (in) Max Temperature (° F) Power (VAC)
Availability Pricing
COMPARE
4 1/2 212 115
$273.75USD / Each
George Ulanet 324-230 Heat-O-Matic Immersion Heater, short Rod , 5" L Heated Area, 1100 watts, 115 VAC EW-03046-04 Mfr # M306-02-00
COMPARE
5 212 115
$264.50USD / Each
COMPARE
3 212 115
$286.25USD / Each
COMPARE
2 3/4 212 115
$296.25USD / Each
George Ulanet 306-6-115-1100 Heat-O-Matic Immersion Heater, Long Rod , 20" L Heated Area, 500 watts, 115 VAC EW-03046-50 Mfr # M306-01-00
COMPARE
18 1/2 212 115
$365.00USD / Each
George Ulanet 306-20"-1100W-230V Heat-O-Matic Immersion Heater, Long Rod , 20" L Heated Area, 1100 watts, 230 VAC EW-03046-56 Mfr # 306-20"-1100W-230V
COMPARE
18 1/2 212 230
$339.50USD / Each