Heavy-Duty Bag Filter Housings
Unique bar-knob closure is safer and faster than clamp designs
  • High-flow housings for flow rates up to 220 GPM
  • Solids are neatly contained for removal with bag changeouts
  • Ideal for commercial, industrial, and process applications
This general-purpose housing design has a closure assembly that works better than most clamp designs: the cover cannot be improperly installed, reducing the chance for cover blowoff if improperly sealed. The seal is positive 100% of the time and only requires two hands to install, not three hands
like many clamp designs. The closures can also be easily replaced if damaged. Housings are height-adjustable within the leg assembly; also included with the housing is a Viton® cover seal and stainless steel filter bag bracket. Maximum temperature for aluminum housings is 300°F (149°C) and 400°F (204°C) for stainless steel.
Common applications include filtration of coolants, solvents, process water, ground water and oils for food, lubrication and hydraulics.
Order filter bags separately.
$1,116.00 - $2,450.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Flow Rate Material Port Size
Availability Pricing
COMPARE
20 GPM Aluminum 0.25" NPT(F)
$1,116.00USD / Each
COMPARE
40 GPM Aluminum 0.25" NPT(F)
$1,306.00USD / Each
COMPARE
90 GPM Aluminum 2" NPT(F)
$1,920.00USD / Each
COMPARE
200 GPM Aluminum 2" NPT(F)
$2,202.00USD / Each
COMPARE
220 GPM Aluminum 3" NPT(F)
$2,450.00USD / Each