McMillan Flow High-Accuracy Stainless Steel Flowmeters and Transmitters
Economical design with an outstanding ±1.0 % full-scale accuracy
  • A wide range of outputs options for integration into any system
Select these flowmeters and transmitters for extremely low flow-rate applications or for higher flow rate measurements and yet maintain a full-scale accuracy of ±1% or better. All units feature a stainless steel housing and come standard with stainless steel compression fittings.
The flowmeters/transmitters can be used with a wide variety of transparent, low-viscosity (under 10 cSt) corrosive or neutral liquids and solvents.
Note: A power supply is required for each meter/ transmitter and may be ordered separately. If using an existing power supply, order cable assembly 32704-52.
$664.50 - $1,041.50USD / Each
21 variations of this product are available.
See All Product Options Below
21 PRODUCT OPTIONS
Compareitem specifications
Display Type Max Flow Rate (mL/min) Output
Availability Pricing
McMillan Flow 104-3 Stainless steel flow sensors with 0 to 5 VDC outputs 13 to 100 mL/min flow rate EW-32718-00 Mfr # 104-3
COMPARE
0 to 5 VDC
$666.50USD / Each
McMillan Flow 104-4 Stainless steel flow sensors with 0 to 5 VDC outputs 20 to 200 mL/min flow rate EW-32718-02 Mfr # 104-4
COMPARE
0 to 5 VDC
$666.50USD / Each
McMillan Flow 104-5 Stainless steel flow sensors with 0 to 5 VDC output; 50 to 500 mL/min flow rate EW-32718-04 Mfr # 104-5
COMPARE
0 to 5 VDC
$677.25USD / Each
McMillan Flow 104-6 Stainless steel flow sensors with 0 to 5 VDC outputs 0.1 to 1 L/min flow rate EW-32718-06 Mfr # 104-6
COMPARE
0 to 5 VDC
$664.50USD / Each
McMillan Flow 104-7 Stainless steel flow sensors with 0 to 5 VDC output; 0.2 to 2 L/min flow rate EW-32718-07 Mfr # 104-7
COMPARE
0 to 5 VDC
$666.50USD / Each
McMillan Flow 104-8 Stainless steel flow sensors with 0 to 5 VDC outputs; 0.5 to 5 L/min flow rate EW-32718-08 Mfr # 104-8
COMPARE
0 to 5 VDC
$734.50USD / Each
McMillan Flow S-114-3 High-Accuracy Stainless Steel Flowmeter, 13 to 100 mL/min EW-32718-20 Mfr # S-114-3
COMPARE
3.5-digit LCD 0 to 5 VDC
$982.75USD / Each
McMillan Flow S-114-4 High-Accuracy Stainless Steel Flowmeter, 20 to 200 mL/min EW-32718-22 Mfr # S-114-4
COMPARE
3.5-digit LCD 0 to 5 VDC
$973.50USD / Each
McMillan Flow S-114-5 High-Accuracy Stainless Steel Flowmeter, 50 to 500 mL/min EW-32718-24 Mfr # S-114-5
COMPARE
3.5-digit LCD 0 to 5 VDC
$955.75USD / Each
McMillan Flow S-114-6 High-Accuracy Stainless Steel Flowmeter, 0.1 to 1 LPM EW-32718-26 Mfr # S-114-6
COMPARE
3.5-digit LCD 0 to 5 VDC
$932.25USD / Each
McMillan Flow S-114-7 High-Accuracy Stainless Steel Flowmeter, 0.2 to 2 LPM EW-32718-28 Mfr # S-114-7
COMPARE
3.5-digit LCD 0 to 5 VDC
$982.75USD / Each
McMillan Flow S-114-8 High-Accuracy Stainless Steel Flowmeter, 0.5 to 5 LPM EW-32718-30 Mfr # S-114-8
COMPARE
3.5-digit LCD 0 to 5 VDC
$1,041.50USD / Each
McMillan Flow S-114-9 High-Accuracy Stainless Steel Flowmeter, 1 to 10 LPM EW-32718-32 Mfr # S-114-9
COMPARE
3.5-digit LCD 0 to 5 VDC
$1,030.00USD / Each
McMillan Flow 107-3 Stainless steel flow sensors with 4 to 20 mA output; 13 to 100 mL/min flow rate EW-32718-40 Mfr # 107-3
COMPARE
4 to 20 mA
$799.00USD / Each
McMillan Flow 107-4 Stainless steel flow sensors with 4 to 20 mA; 20 to 200 mL/min flow rate EW-32718-42 Mfr # 107-4
COMPARE
4 to 20 mA
$783.00USD / Each
McMillan Flow 107-5 Stainless steel flow sensors with 4 to 20 mA output; 50 to 500 mL/min flow rate EW-32718-44 Mfr # 107-5
COMPARE
4 to 20 mA
$772.50USD / Each
McMillan Flow 107-6 Stainless steel flow sensors with 4 to 20 mA output; 0.1 to 1 L/min flow rate EW-32718-46 Mfr # 107-6
COMPARE
4 to 20 mA
$783.00USD / Each
McMillan Flow 107-7 Stainless steel flow sensors with 4 to 20 mA output; 0.2 to 2 L/min flow rate EW-32718-48 Mfr # 107-7
COMPARE
4 to 20 mA
$668.50USD / Each
McMillan Flow 107-8 Stainless steel flow sensors with 4 to 20 mA output; 0.5 to 5 L/min flow rate EW-32718-50 Mfr # 107-8
COMPARE
4 to 20 mA
$843.75USD / Each
McMillan Flow 107-9 Stainless steel flow sensors with 4 to 20 mA output; 1 to 10 L/min flow rate EW-32718-52 Mfr # 107-9
COMPARE
4 to 20 mA
$964.25USD / Each
Showing 1 - 20 of 21