High-Flow Polyester Felt-Filter Bags
Wide range of options for temperature, flow rate, and filtration quality
  • Constructed of Polyester Felt
Designed for higher flow with an average efficiency at the rated pore size. Polyester felt is ideal for general-purpose filtration. The bags are glazed to minimize fiber migration. Choose polyester for higher temperature applications, rated for a temperature range of 300°F (149°C) maximum.
$4.50 - $5.50USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Filter Media Flow Rate Micron Rating
Availability Pricing
COMPARE
Polyester 20 GPM 100
$4.50USD / Each
COMPARE
Polyester 40 GPM 10
$5.50USD / Each
COMPARE
Polyester 40 GPM 25
$5.50USD / Each