Hoffman A51FL Continuous-Hinge Steel Junction Boxes with Quarter-Turn Fast Latch, Type 4, 12
This enclosure provides the convenience of quarter-turn latching with an aesthetic, easy-to-maintain light gray textured finish
  • Quarter-turn latch with screwdriver slot for easy access
  • Oil-resistant, seamless foam-in-place gasket
  • Available in 14 or 16 gauge steel
  • Seams continuously welded and ground smooth
  • Continuous hinge
  • Bonding provision on door
  • Weld nuts provided for mounting optional panels and terminal block kits in size 6” x 4” (152 x 102 mm) and larger
  • Ratings: NEMA Type 4, 12; IP66
11 PRODUCT OPTIONS
Compareitem specifications
Outside Depth (in) Outside Height (in) Outside Width (in)
Availability Pricing
Pentair Hoffman A16106CHFL Type 4 Quarter-Turn Fast Latch Steel Junction Box EW-10501-80 Mfr # A16106CHFL
COMPARE
$252.00USD / Each
Pentair Hoffman A10086CHFL Type 4 Quarter-Turn Fast Latch Steel Junction Box EW-10502-58 Mfr # A10086CHFL
COMPARE
$180.00USD / Each
Pentair Hoffman A1008CHFL Type 4 Quarter-Turn Fast Latch Steel Junction Box EW-10502-59 Mfr # A1008CHFL
COMPARE
$155.00USD / Each
Pentair Hoffman A12108CHFL Type 4 Quarter-Turn Fast Latch Steel Junction Box EW-10502-60 Mfr # A12108CHFL
COMPARE
$240.00USD / Each
Pentair Hoffman A1210CHFL Type 4 Quarter-Turn Fast Latch Steel Junction Box EW-10502-61 Mfr # A1210CHFL
COMPARE
$198.00USD / Each
Pentair Hoffman A1412CHFL Type 4 Quarter-Turn Fast Latch Steel Junction Box EW-10502-62 Mfr # A1412CHFL
COMPARE
$250.00USD / Each
Pentair Hoffman A1614CHFL Type 4 Quarter-Turn Fast Latch Steel Junction Box EW-10502-63 Mfr # A1614CHFL
COMPARE
$294.00USD / Each
Pentair Hoffman A606CHFL Type 4 Quarter-Turn Fast Latch Steel Junction Box EW-10502-64 Mfr # A606CHFL
COMPARE
$118.00USD / Each
Pentair Hoffman A8066CHFL Type 4 Quarter-Turn Fast Latch Steel Junction Box EW-10502-65 Mfr # A8066CHFL
COMPARE
$145.00USD / Each
Pentair Hoffman A806CHFL Type 4 Quarter-Turn Fast Latch Steel Junction Box EW-10502-66 Mfr # A806CHFL
COMPARE
$129.00USD / Each
Pentair Hoffman A808CHFL Type 4 Quarter-Turn Fast Latch Steel Junction Box EW-10502-67 Mfr # A808CHFL
COMPARE
$140.00USD / Each
Showing 1- 11 of 11
Need Help with this product?
Call Us At1-847-549-7600
Live chat