Hollow Polypropylene (PP) Ball Bath Covers
Reduce vapor and heat loss by 75% with one layer of balls
Polypropylene balls form a tight cover over liquids to prevent change in temperature and reduce evaporation. Use instead of a bath lid with any of the baths listed in the section.
$133.00 - $566.00USD / Pkg of 2000
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Hollow PolyPropylene (PP) Ball Bath Covers, 10 mm; 2500/Pk
$271.00USD / Pkg of 2500
COMPARE
Hollow Polypropylene (PP) Ball Bath Covers, 20 mm; 500/Pk
$133.00USD / Pkg of 500
COMPARE
Hollow Polypropylene (PP) Ball Bath Covers, 20 mm; 1000/Pk
$183.00USD / Pkg of 1000
COMPARE
Hollow Polypropylene (PP) Ball Bath Covers, 38 mm; 500/Pk
$306.00USD / Pkg of 500
COMPARE
Hollow Polypropylene (PP) Ball Bath Covers, 38 mm; 1000/Pk
$566.00USD / Pkg of 1000
COMPARE
Hollow Polypropylene (PP) Ball Bath Covers, 10 mm, 5000/pk
$465.00USD / Pkg of 5000
COMPARE
Hollow Polypropylene (PP) Ball Bath Covers, 20 mm, 2000/pk
$224.00USD / Pkg of 2000
COMPARE
Hollow Polypropylene (PP) Ball Bath Covers, 38 mm, 1000/pk
$556.00USD / Pkg of 1000
Showing 1- 8 of 8