Honeywell CoreShield™ Gloves

Sturdy cut-resistant gloves simplify selection for a variety of applications
  • Color-coded marking system highlights the right gloves for your application, per EN/ANSI standards
  • Choose by level of cut resistance, protection, grip, dexterity and fit
  • One of the lightest-weight cut-protective gloves on the market
  • Touchscreen resistive and capacitive
  • Water and oil proof coating
Providing durability and dexterity, value-packed CoreShield™ gloves from Honeywell offer optimum cut protection and user comfort. The lightweight gloves prevent hand fatigue, while a specially designed microfoam elastic coating provides outstanding oil- and dry-grip performance.

Each glove is made with softer, skin-friendly yarns that--compared to glass fiber--reduce the allergy risk to wearers. Due to this, all CoreShield™ gloves are certified STANDARD 100 by OEKO-TEX®.

CoreShield™ coatings offer 360° breathability through an open cell structure, further enhancing comfort even upon extensive wear. The washability of the glove material means little change in performance or size after five washes.
$18.20 - $18.70USD / Pair of 1

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Size Description
Availability Pricing
Honeywell Coreshield™ 10 Gauge ANSI A9/F Glove, 7/S
Buy More Save More
EW-86326-64 Mfr # 29-0910B/7S
COMPARE
7/S Honeywell Coreshield™ 10 Gauge ANSI A9/F Glove, 7/S
$18.20USD / Pair of 1
Honeywell Coreshield™ 10 Gauge ANSI A9/F Glove, 8/M
Buy More Save More
EW-86326-65 Mfr # 29-0910B/8M
COMPARE
8/M Honeywell Coreshield™ 10 Gauge ANSI A9/F Glove, 8/M
$18.70USD / Pair of 1
Honeywell Coreshield™ 10 Gauge ANSI A9/F Glove, 9/L
Buy More Save More
EW-86326-66 Mfr # 29-0910B/9L
COMPARE
9/L Honeywell Coreshield™ 10 Gauge ANSI A9/F Glove, 9/L
$18.70USD / Pair of 1
Honeywell Coreshield™ 10 Gauge ANSI A9/F Glove, 10/XL
Buy More Save More
EW-86326-67 Mfr # 29-0910B/10XL
COMPARE
10/XL Honeywell Coreshield™ 10 Gauge ANSI A9/F Glove, 10/XL
$18.70USD / Pair of 1
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat