This product is now discontinued, View Related Products
Honeywell Laser Eyewear, XC Series
Excellent visible light transmission minimizes distortion
These lightweight, impact-resistant polymer lenses are suitable for low-powered lasers or indirect laser exposure. Wavelength, optical density, and lot number are inscribed across lens for easy identification. All eyewear comes with protective case, cleaning cloth, and safety hanging cord.
XC Series
The industry's first 9-base wraparound laser eyewear encompasses the eye while theunique Multi-Material Technology® (MMT) cushions every point of contact with the face for unparalleled comfort and impact resistance. Cushioned temples with soft, flexible nose fingers adjust to fit a variety of facial profiles. Lens and temple adjustability options further customize the fit. Detachable prescription insert is included with every pair.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Laser Type Lens Color
Availability Pricing
Honeywell 31-80137 Laser Safety Glasses, YAG/CO2 EW-86438-20 Mfr # 31-80137
COMPARE
YAG, CO2 Green
Call For Price
Honeywell 31-80108 Laser Safety Glasses, YAG/Diode EW-86438-21 Mfr # 31-80108
COMPARE
YAG, diode Green
Call For Price
Honeywell 31-80102 XC Laser Eyewear, YAG/Harmonics EW-86438-22 Mfr # 31-80102
COMPARE
YAG, harmonics Brown