Horiba Oil Content Analyzer Solvent Reclaimer
Save money on solvent costs
  • Reduce overall volume of solvents used and discarded
Designed especially for reclaiming S-316 solvent, this unit features a double column of activated carbon and activated aluminum. It has a large filtering capacity, is easy to operate, and requires no electricity. Highly efficient, the SR-305 can reduce your per test solvent cost.
$955.50 - $3,547.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Horiba 100690 Extraction Solvent S-316 for Oil Content Analyzer; 1.5 kg
Always in Stock
EW-82900-10 Mfr # 5200100690
COMPARE
Solvent extractant S-316, 1.5 kg, for Oil Content Analyzer
$955.50USD / Each
Horiba SR-305 Solvent Reclaimer for S-316 Extraction Solvent EW-82900-20 Mfr # 3014102236
COMPARE
Solvent reclaimer SR-305, for recycling Oil Content Analyzer extractant
$3,547.00USD / Each