Hose Barb Elbow Inserts 57161
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting—fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.
$6.80 - $211.00USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Fitting Material
Availability Pricing
CPC (Colder) PMC2302 Quick-Disconnect Hose Barb Elbow Insert; Straight-through, Delrin, 1/8" Tube ID
Buy More Save More
EW-06361-27 Mfr # PMC2302
COMPARE
1/8 Acetal
$6.80USD / Each
CPC (Colder) PMC2304 Hose Barb Elbow Insert; Straight-through, Delrin, 1/4" Tube ID, 1/8" Flow Size
Buy More Save More
EW-06361-28 Mfr # PMC2304
COMPARE
1/4 Acetal
$6.80USD / Each
CPC (Colder) PMCD2302 Quick-Disconnect Hose Barb Elbow Insert; Valved, Delrin, 1/8" Tube ID
Buy More Save More
EW-06361-39 Mfr # PMCD2302
COMPARE
1/8 Acetal
$13.70USD / Each
CPC (Colder) PMCD2304 Quick-Disconnect Hose Barb Elbow Insert; Valved, Delrin, 1/4" Tube ID, 1/8" Flow Size
Buy More Save More
EW-06361-41 Mfr # PMCD2304
COMPARE
1/4 Acetal
$13.90USD / Each
CPC (Colder) PMC230212 Quick-Disconnect Hose Barb Elbow Insert; Straight-through, PP, 1/8" Tube ID
Buy More Save More
EW-06361-85 Mfr # PMC230212
COMPARE
1/8 Polypropylene
$9.40USD / Each
CPC (Colder) PMCD230212 Quick-Disconnect Hose Barb Elbow Insert; Valved, PP, 1/8" Tube ID
Buy More Save More
EW-06361-98 Mfr # PMCD230212
COMPARE
1/8 Polypropylene
$18.00USD / Each
CPC (Colder) PMCD230412 Quick-Disconnect Hose Barb Elbow Insert; Valved, PP, 1/4" Tube ID, 1/8" Flow Size
Buy More Save More
EW-06361-99 Mfr # PMCD230412
COMPARE
1/4 Polypropylene
$15.80USD / Each
CPC (Colder) PMC230412 Quick-Disconnect Hose Barb Elbow Insert; Straight-through, PP, 1/4" Tube ID; 25/PK EW-31307-67 Mfr # PMC230412
COMPARE
1/4 Polypropylene
$211.00USD / Pkg of 25
Showing 1- 8 of 8