This product is now discontinued, View Related Products

Huber Unistat Tango Heating/Cooling Recirculators

Atmospherically sealed thermostat can be set above or below the level of the application
  • Environmentally friendly temperature control units utilize natural refrigerants
  • Electronic, user-adjustable overtemperature protection and optical low-level sensor
  • Intuitive Pilot ONE touchscreen controller simplifies tasks
These heating/cooling circulators use environmentally friendly, natural refrigerants while boasting a powerful, variable-speed pressure/suction pump—ideal for either open or closed-loop applications.
The integrated Pilot ONE controller features a large 5.7" (14.5 cm), color touch-screen display with 11 language support (EN, DE, FR, IT, ES, RU, CN, PT, JP, CZ, and PL), convenient color-coded navigation system, and USB and Ethernet ports for communication and remote control through your tablet, PC, or network. Additional controller capabilities provide further functionality and include programmability (10 programs, 100 steps each), ramping (linear and nonlinear), five-point calibration, and the option to control the system based on the internal or process temperature.
Call For Price

6 variations of this product are available.

See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Power (Hz) Power (VAC) Description
Availability Pricing
Huber Unistat Tango Heating/Cooling Recirculator, Air Cooled; 208VAC/60Hz EW-12125-84 Mfr # 1000.0018.01
COMPARE
60 208 Heating/Cooling Recirculator, Air Cooled; 208VAC/60Hz
Call For Price
Huber Unistat Tango Heating/Cooling Recirculator, Air/Water Cooled; 208VAC/60Hz EW-12125-85 Mfr # 1000.0024.01
COMPARE
60 208 Heating/Cooling Recirculator, Air/Water Cooled; 208VAC/60Hz
Call For Price
Huber Unistat Tango Heating/Cooling Recirculator, Water Cooled; 208VAC/60Hz EW-12125-86 Mfr # 1000.0023.01
COMPARE
60 208 Heating/Cooling Recirculator, Water Cooled; 208VAC/60Hz
Call For Price
Huber Unistat Tango Heating/Cooling Recirculator, Air Cooled; 230VAC/50Hz EW-12125-87 Mfr # 1000.0016.01
COMPARE
50 230 Heating/Cooling Recirculator, Air Cooled; 230VAC/50Hz
Call For Price
Huber Unistat Tango Heating/Cooling Recirculator, Air/Water Cooled; 230VAC/50Hz EW-12125-88 Mfr # 1000.0017.01
COMPARE
50 230 Heating/Cooling Recirculator, Air/Water Cooled; 230VAC/50Hz
Call For Price
Huber Unistat Tango Heating/Cooling Recirculator, Water Cooled; 230VAC/50Hz EW-12125-89 Mfr # 1000.0021.01
COMPARE
50 230 Heating/Cooling Recirculator, Water Cooled; 230VAC/50Hz
Call For Price
Showing 1- 6 of 6

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat