This product is now discontinued, View Related Products
HybriLinker Hybridization Incubator
Hybridizer and UV crosslinker in one self-contained space-saving unit
  • Each section able to operate at the same time
  • Precise temperature control, ambient to 99.9°C and variable speed control at 10 to 18 rpm
  • Multiple bottle sizes fits up to ten with three different sized bottle configurations
The microprocessor-controlled hybridization oven monitors temperature and rotation speed of bottles for dynamic mixing capability. This is ideal for creating the optimum conditions needed for cDNA library screenings, primer synthesis and all nucleic acid hybridization. The UV crosslinker provides an extremely uniform shortwave radiation exposure chamber for bonding nucleic acid to medium, typically done prior to hybridization. UV output is measured by an internal sensor and controlled by an on-board microprocessor, enabling accurate, reliable and safe laboratory crosslinking. Preset or user-selected UV energy or UV time exposure are selected via touch pad.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
Hybrilinker Hybridization Incubator, 115v/60Hz EW-39453-14 Mfr # 95-0031-01
COMPARE
115
Call For Price
Hybrilinker Hybridization Incubator, 230v/50Hz EW-39453-16 Mfr # 95-0031-02
COMPARE
230