Hydraulically Actuated Diaphragm Pumps 58366
High suction lift and high viscosity combine in the stainless steel pump
  • Achieve up to 65 GPH or 1100 psi
  • Internal bypass allows for improved valve performance
  • Pump capacity is adjustable through 100% of range using a manual micrometer dial/automatic electric positioner with manual override
Pump drive components are fully enclosed and operate in an oil bath to minimize friction and wear,
as well as to protect from harsh environments. Pump provides metering accuracy of ±1%. A micrometer
knob enables capacity adjustment between 0 to 100% whether the pump is operating or stopped.
$2,101.00 - $2,878.50USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (GPH) Max Pressure (PSI) Wetted Materials
Availability Pricing
Neptune 515-E-N3 Hydraulically Actuated Diaphragm Pump; SS/PTFE, 3.0 GPH at 1100 PSI EW-74151-00 Mfr # 515-E-N3
COMPARE
1100 316 SS, PTFE
$2,153.50USD / Each
Neptune 515-E-N5 Hydraulically Actuated Diaphragm Pump, PVC and PTFE Wetted End, 3.0GPH @ 150PSI EW-74151-02 Mfr # 515-E-N5
COMPARE
150 Glass, PTFE, PVC
$2,133.50USD / Each
Neptune 525-E-N3 Hydraulically Actuated Diaphragm Pump; SS/PTFE, 7.0 GPH at 900 PSI EW-74151-10 Mfr # 525-E-N3
COMPARE
900 316 SS, PTFE
$2,101.00USD / Each
Neptune 532-E-N3 Hydraulically Actuated Diaphragm Pump; SS/PTFE, 11 GPH at 350 PSI EW-74151-20 Mfr # 532-E-N3
COMPARE
350 316 SS, PTFE
$2,143.50USD / Each
Neptune 532-E-N5 Hydraulically Actuated Diaphragm Pump, PVC and PTFE Wetted End, 11.0GPH @ 150PSI EW-74151-22 Mfr # 532-E-N5
COMPARE
150 Glass, PTFE, PVC
$2,285.00USD / Each
Neptune 535-E-N3 Hydraulically Actuated Diaphragm Pump; SS/PTFE, 18 GPH at 350 PSI EW-74151-30 Mfr # 535-E-N3
COMPARE
350 316 SS, PTFE
$2,143.50USD / Each
Neptune 547-SE-N3 Hydraulically Actuated Diaphragm Pump; SS/PTFE, 30 GPH at 200 PSI EW-74151-40 Mfr # 547-SE-N3
COMPARE
200 316 SS, PTFE
$2,315.50USD / Each
Neptune 547-S-N5 Hydraulically Actuated Diaphragm Pump, PVC and PTFE Wetted End, 30.0GPH @ 300PSI EW-74151-42 Mfr # 547-S-N5
COMPARE
300 Glass, PTFE, PVC
$2,878.50USD / Each
Showing 1- 8 of 8