Hydrogen (H2) Gas Mass Flowmeters with Metal bodies
Precisely measure flow rates and totals
  • Proven design uses metal body components for added durability
  • Accumulated flow can be logged using an optional totalizer
  • Includes NIST-traceable certificate
These flowmeters feature an advanced straight-tube sensor that ensures accurate and repeatable results. Gas flow measurements are unaffected by moderate temperature and pressure variations at the inlet. The meter also allows a four-point calibration across the flow range to improve meter linearity. Output data from the meter can be sent via a 0 to 5 VDC or 4 to 20 mA signal; an analog to-RS converter is available for data collection and analysis on your PC.
All meters include a detachable 3 1/2-digit LCD that can be tilted up to 90° for easy reading. Remotely mount the display up to three feet using extension cable 32662-70, cable sold separately. The optional totalizer 32650-22 displays accumulated flow. The meter is protected from polarity reversal or short circuit by a built-in resettable fuse. Aluminum models have wetted materials of anodized aluminum, brass, and Viton®; stainless steel (SS) models have wetted materials of SS and Viton.
These precalibrated flow controllers operate at inlet pressures between 5 and 60 psi and at gas temperatures between 59 and 77°F (15 to 25°C) while maintaining the stated percent full-scale accuracy and linearity. When operating beyond 5 to 60 psi, add ±0.01%/psi full-scale; if operating beyond 59 to 77°F (15 to 25°C), add ±0.15%/°C full-scale.
Power supply allows flowmeters to be powered by AC voltage. Order 110 VAC power supply 03277-00, or 220 VAC power supply 03277-05. Power supplies are sold separately.
$650.00 - $1,398.00USD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (LPM) Process Connection Wetted Materials
Availability Pricing
Aalborg Thermal Gas Mass Flowmeter, to 20 sccm H2, Aluminum body w/ display
Buy More Save More
EW-32654-02 Mfr # GFMS-010133
COMPARE
0.02 1/4" Compression Fittings Anodized Aluminum, Brass, Viton
$650.00USD / Each
Aalborg Thermal Gas Mass Flowmeter, to 200 sccm H2, Aluminum body w/ display
Buy More Save More
EW-32654-08 Mfr # GFMS-010136
COMPARE
0.2 1/4" Compression Fittings Anodized Aluminum, Brass, Viton
$650.00USD / Each
Aalborg Thermal Gas Mass Flowmeter, to 500 sccm H2, Aluminum body w/ display
Buy More Save More
EW-32654-10 Mfr # GFMS-010364
COMPARE
0.5 1/4" Compression Fittings Anodized Aluminum, Brass, Viton
$650.00USD / Each
Aalborg Thermal Gas Mass Flowmeter, to 1 L/min H2, Aluminum body w/ display
Buy More Save More
EW-32654-12 Mfr # GFMS-010138
COMPARE
1 1/4" Compression Fittings Anodized Aluminum, Brass, Viton
$650.00USD / Each
Aalborg Thermal Gas Mass Flowmeter, to 5 L/min H2, Aluminum body w/ display
Buy More Save More
EW-32654-16 Mfr # GFMS-010140
COMPARE
5 1/4" Compression Fittings Anodized Aluminum, Brass, Viton
$650.00USD / Each
Aalborg Thermal Gas Mass Flowmeter, to 15 L/min H2, Aluminum body w/ display
Buy More Save More
EW-32654-19 Mfr # GFM371-11/H2
COMPARE
15 1/4" Compression Fittings Anodized Aluminum, Brass, Viton
$814.00USD / Each
Aalborg GFMS Thermal Gas Mass Flowmeter with Display, 500 sccm H2, 316SS Body
Buy More Save More
EW-32654-60 Mfr # GFMS-010152
COMPARE
0.5 1/4" Compression Fittings 316 Stainless Steel, Viton
$984.00USD / Each
Aalborg GFMS Thermal Gas Mass Flowmeter with Display, 5 L/min H2, 316SS Body
Buy More Save More
EW-32654-66 Mfr # GFMS-010155
COMPARE
5 1/4" Compression Fittings 316 Stainless Steel, Viton
$984.00USD / Each
Aalborg GFMS Thermal Gas Mass Flowmeter with Display, 50 L/min H2, 316SS Body
Buy More Save More
EW-32654-86 Mfr # GFMS-010158
COMPARE
50 1/4" Compression Fittings 316 Stainless Steel, Viton
$1,222.00USD / Each
Aalborg Thermal Gas Mass Flowmeter, to 100 L/min H2, 316SS body w/ display
Buy More Save More
EW-32654-92 Mfr # GFMS-010159
COMPARE
100 3/8" Compression Fittings 316 Stainless Steel, Viton
$1,398.00USD / Each
Showing 1- 10 of 10