I/P Precision Modular Pump Systems
Modular drive lets you place motor and controller where convenient—ideal for limited or awkward spaces
  • Minimum flow rate is 0.06 LPM (0.02 GPM); maximum flow rate is 8 LPM (2.1 GPM)
  • Three-digit LED display shows motor rpm—confirm speed setting at a glance
  • Membrane keypad for easy up-front control of speed, direction, and start/stop
  • Reversible motor—purge tubing before or after pumping; pump fluid in either direction
  • LED indicators confirm motor direction
  • Powerful motors let you stack two pump heads for increased flow rates and for multichannel applications
  • Separate speed and power (on/off) control—maintain speed setting while turning pump on/off
  • ±0.25% speed control; meter and transfer fluids with accuracy and repeatability
  • Remote control capabilities via DB9 female connector on back of drive: Speed control input 4 to 20 mA or 0 to 10 V; Pump direction (CW/CCW) via open collector or contact closure; Start/stop via open collector or contact closure
  • Splash-resistant ABS plastic controller housing is IP33 rated; drive housing is epoxy finished steel, IP33 rated
  • 6-ft (1.8-m) cable connects drive and controller; order optional extension cable 07557-51 to extend to 15 ft (4.6 m)
I/P® modular drives feature reversible pumping and allow you the flexibility to separate the motor from the controller. These accurate modular drives have powerful motors, speed display, and ±0.25% speed control for precise flow regulation—ideal for metering and critical transfer applications.
$3,470.00 - $4,340.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Drive Drive IP rating Washdown Capable
Availability Pricing
COMPARE
07591-20 (I/P® Variable-Speed Wash-Down Modular Drive) IP 33 No
$3,470.00USD / Each
COMPARE
07591-30 (I/P® Variable-Speed Wash-Down Modular Drive) IP 66 Yes
$4,340.00USD / Each