ICI Quick Ship Drop-In Epoxy Resin Sinks
A sink for every lab setup
  • Chemical-resistant epoxy construction
  • Easily convert into undermount style
  • ADA-compliant option
Black epoxy resin drop-in sinks are chemically resistant, feature coved corners, pitch to a center or corner drain, and are furnished with a tail piece and strainer. Model 78985-29 is an ADA-compliant option.
$1,125.00 - $1,940.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Depth (in) Length (in) Width (in)
Availability Pricing
ICI Quick Ship Drop-In Sink, Epoxy, 18" L, 15" W, 11" D, Corner Outlet EW-78985-25 Mfr # CPD-30
COMPARE
11 18 15
$1,125.00USD / Each
ICI Quick Ship Drop-In Sink, Epoxy, 24" L, 16" W, 8" D, Center Outlet EW-78985-26 Mfr # CPD-50
COMPARE
8 24 16
$1,125.00USD / Each
ICI Quick Ship Drop-In Sink, Epoxy, 25" L, 15" W, 10" D, Corner Outlet EW-78985-27 Mfr # CPD-55
COMPARE
10 25 15
$1,225.00USD / Each
ICI Quick Ship Drop-In Sink, Epoxy, 28" L, 15" W, 11.8" D, Corner Outlet EW-78985-28 Mfr # CPD-59
COMPARE
11 51/64 28 15
$1,940.00USD / Each
ICI Quick Ship ADA Drop-In Sink, Epoxy, 25" L, 15" W, 4.8" D, Corner Outlet EW-78985-29 Mfr # CPA-55
COMPARE
4 51/64 25 15
$1,200.00USD / Each