ICI Quick Ship Electrical Pedestals
Electrical pedestals for a quick delivery furniture program that builds complete labs
  • An extensive inventory of high quality furnishings, chemical fume hoods, and components
  • Clear and easy project communication
  • Design, project management, and factory trained installation
Electrical pedestals come in a single gang and double gang. The power supplied is 120 V, 20 amp. The outlets are black with stainless steel cover plates while the box is cast aluminum and powder coated with a black, textured epoxy finish.
A custom electrical pedestal is also available to accommodate a variety of styles and configurations. The custom pedestal is double faced and as many as four duplex outlets can be installed. The chambered design reduces RF interference when electrical and data outlets are combined in this pedestal. The box is cast aluminum and powder coated with a black, textured epoxy finish. Outlets and cover plates are sold separately.
$185.00 - $624.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
ICI Quick Ship Single Gang, Double Faced Electrical Pedestal, Standard Outlets EW-78985-38 Mfr # CPE433WS
COMPARE
Single Gang, Double Faced Electrical Pedestal, Standard Outlets
$185.00USD / Each
ICI Quick Ship Single Gang, Double Faced Electrical Pedestal, GFI Outlets EW-78985-39 Mfr # CPE433WS-GFI
COMPARE
Single Gang, Double Faced Electrical Pedestal, GFI Outlets
$364.00USD / Each
ICI Quick Ship Double Gang, Double Faced Electrical Pedestal, Standard Outlets EW-78985-40 Mfr # CPE633WS
COMPARE
Double Gang, Double Faced Electrical Pedestal, Standard Outlets
$265.00USD / Each
ICI Quick Ship Double Gang, Double Faced Electrical Pedestal, GFI Outlets EW-78985-41 Mfr # CPE633WS-GFI
COMPARE
Double Gang, Double Faced Electrical Pedestal, GFI Outlets
$624.00USD / Each
ICI Quick Ship Double Gang, Custom Electrical/Data Pedestal EW-78985-42 Mfr # CPE800WS
COMPARE
Double Gang, Custom Electrical/Data Pedestal
$554.00USD / Each