ICI Quick Ship Exhaust Snorkels
Safely eliminates heat and harmful fumes
  • Designed to be mounted from the wall or ceiling
  • Arm joints allow snorkel to rotate and swivel 360°
These lightweight, flexible exhaust snorkels with three adjustable, composite friction joints allow you to get close to the source of fumes and particles safely. The two outermost arm joints can rotate and swivel 360 degrees. Constructed of anodized aluminum, the snorkels offer strength, durability, and ease of handling. The arms are 3” in diameter and are available with a maximum working radius of 42.9” (109 cm) or 57.1” (145 cm). Each arm comes with a 5.5” mini hood that will accept optional hoods.
$2,329.00 - $2,967.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
ICI Quick Ship Original Exhaust Snorkel, 42.9" (109 cm) Radius EW-78981-14 Mfr # CPOES-3
COMPARE
ICI Quick Ship Original Exhaust Snorkel, 42.9" (109 cm) Radius
$2,329.00USD / Each
ICI Quick Ship Original Exhaust Snorkel, 57.1"  (145 cm) Radius EW-78981-15 Mfr # CPOES-3L
COMPARE
ICI Quick Ship Original Exhaust Snorkel, 57.1" (145 cm) Radius
$2,662.00USD / Each
ICI Quick Ship CHEM Exhaust Snorkel, 42.9" (109 cm) Radius EW-78981-16 Mfr # CPCES-3
COMPARE
ICI Quick Ship CHEM Exhaust Snorkel, 42.9" (109 cm) Radius
$2,667.00USD / Each
ICI Quick Ship CHEM Exhaust Snorkel, 57.1"  (145 cm) Radius EW-78981-17 Mfr # CPCES-3L
COMPARE
ICI Quick Ship CHEM Exhaust Snorkel, 57.1" (145 cm) Radius
$2,967.00USD / Each