ICI Quick Ship Laboratory Service Valves
Deliver flow control of laboratory gases to your ICI Quick Ship lab benches
  • Designed with deck-mounted turret base for easy flow control
  • Color-coded handles allow for easy gas identification
  • Choose from a wide range of configuration options suitable for any laboratory
Laboratory service valves provide flow control of laboratory gases to your ICI Quick Ship lab setup. These brass service valves are available in both ball and needle valve options. Ball valves provide simple on/off control with a quarter turn of the knob. Needle valves allow for more gradual and precise control with proportional control of the knob. Both valve types are available in four flow pattern configurations—single, two at 90°, two at 180°, and four turrets at 90°. Choose from four service options including gas, air, vacuum, and nitrogen
$168.00 - $722.00USD / Each
32 variations of this product are available.
See All Product Options Below
32 PRODUCT OPTIONS
Compareitem specifications
Description Flow Pattern Wetted Materials
Availability Pricing
ICI Quick Ship Laboratory Ball Valve, One Turret, Gas EW-78984-79 Mfr # CPL4200-131WSA-G
COMPARE
Laboratory Ball Valve, One Turret, Gas One Turret Chrome plated brass valve, forged brass lever
$168.00USD / Each
ICI Quick Ship Laboratory Ball Valve, One Turret, Air EW-78984-80 Mfr # CPL4200-131WSA-A
COMPARE
Laboratory Ball Valve, One Turret, Air One Turret Chrome plated brass valve, forged brass lever
$168.00USD / Each
ICI Quick Ship Laboratory Ball Valve, One Turret, Nitrogen EW-78984-81 Mfr # CPL4200-131WSA-N
COMPARE
Laboratory Ball Valve, One Turret, Nitrogen One Turret Chrome plated brass valve, forged brass lever
$168.00USD / Each
ICI Quick Ship Laboratory Ball Valve, One Turret, Vacuum EW-78984-82 Mfr # CPL4200-131WSA-V
COMPARE
Laboratory Ball Valve, One Turret, Vacuum One Turret Chrome plated brass valve, forged brass lever
$168.00USD / Each
ICI Quick Ship Laboratory Ball Valve, Two 90° Turrets, Gas EW-78984-83 Mfr # CPL4200-132AWSA-G
COMPARE
Laboratory Ball Valve, Two 90° Turrets, Gas Two 90° Turrets Chrome plated brass valve, forged brass lever
$261.00USD / Each
ICI Quick Ship Laboratory Ball Valve, Two 90° Turrets, Air EW-78984-84 Mfr # CPL4200-132AWSA-A
COMPARE
Laboratory Ball Valve, Two 90° Turrets, Air Two 90° Turrets Chrome plated brass valve, forged brass lever
$261.00USD / Each
ICI Quick Ship Laboratory Ball Valve, Two 90° Turrets, Vacuum EW-78984-86 Mfr # CPL4200-132AWSA-V
COMPARE
Laboratory Ball Valve, Two 90° Turrets, Vacuum Two 90° Turrets Chrome plated brass valve, forged brass lever
$261.00USD / Each
ICI Quick Ship Laboratory Ball Valve, Two 180° Turrets, Gas EW-78984-87 Mfr # CPL4200-132SWSA-G
COMPARE
Laboratory Ball Valve, Two 180° Turrets, Gas Two 180° Turrets Chrome plated brass valve, forged brass lever
$261.00USD / Each
ICI Quick Ship Laboratory Ball Valve, Two 180° Turrets, Nitrogen EW-78984-89 Mfr # CPL4200-132SWSA-N
COMPARE
Laboratory Ball Valve, Two 180° Turrets, Nitrogen Two 180° Turrets Chrome plated brass valve, forged brass lever
$261.00USD / Each
ICI Quick Ship Laboratory Ball Valve, Four Turrets, Nitrogen EW-78984-93 Mfr # CPL4200-134WSA-N
COMPARE
Laboratory Ball Valve, Four Turrets, Nitrogen Four 90° Turrets Chrome plated brass valve, forged brass lever
$448.00USD / Each
ICI Quick Ship Laboratory Ball Valve, Four Turrets, Vacuum EW-78984-94 Mfr # CPL4200-134WSA-V
COMPARE
Laboratory Ball Valve, Four Turrets, Vacuum Four 90° Turrets Chrome plated brass valve, forged brass lever
$448.00USD / Each
ICI Quick Ship Laboratory Needle Valve, One Turret, Gas EW-78984-95 Mfr # CPL2880-131WSA-G
COMPARE
Laboratory Needle Valve, One Turret, Gas One Turret Forged brass valve, floating stainless steel needle, replaceable stainless steel seat
$237.00USD / Each
ICI Quick Ship Laboratory Needle Valve, One Turret, Air EW-78984-96 Mfr # CPL2880-131WSA-A
COMPARE
Laboratory Needle Valve, One Turret, Air One Turret Forged brass valve, floating stainless steel needle, replaceable stainless steel seat
$237.00USD / Each
ICI Quick Ship Laboratory Needle Valve, One Turret, Vacuum EW-78984-98 Mfr # CPL2880-131WSA-V
COMPARE
Laboratory Needle Valve, One Turret, Vacuum One Turret Forged brass valve, floating stainless steel needle, replaceable stainless steel seat
$237.00USD / Each
ICI Quick Ship Laboratory Needle Valve, Two 90° Turrets, Gas EW-78984-99 Mfr # CPL2880-132AWSA-G
COMPARE
Laboratory Needle Valve, Two 90° Turrets, Gas Two 90° Turrets Forged brass valve, floating stainless steel needle, replaceable stainless steel seat
$399.00USD / Each
ICI Quick Ship Laboratory Needle Valve, Two 90° Turrets, Vacuum EW-78985-02 Mfr # CPL2880-132AWSA-V
COMPARE
Laboratory Needle Valve, Two 90° Turrets, Vacuum Two 90° Turrets Forged brass valve, floating stainless steel needle, replaceable stainless steel seat
$399.00USD / Each
ICI Quick Ship Laboratory Needle Valve, Two 180° Turrets, Gas EW-78985-03 Mfr # CPL2880-132SWSA-G
COMPARE
Laboratory Needle Valve, Two 180° Turrets, Gas Two 180° Turrets Forged brass valve, floating stainless steel needle, replaceable stainless steel seat
$399.00USD / Each
ICI Quick Ship Laboratory Needle Valve, Two 180° Turrets, Air EW-78985-04 Mfr # CPL2880-132SWSA-A
COMPARE
Laboratory Needle Valve, Two 180° Turrets, Air Two 180° Turrets Forged brass valve, floating stainless steel needle, replaceable stainless steel seat
$399.00USD / Each
ICI Quick Ship Laboratory Needle Valve, Two 180° Turrets, Vacuum EW-78985-06 Mfr # CPL2880-132SWSA-V
COMPARE
Laboratory Needle Valve, Two 180° Turrets, Vacuum Two 180° Turrets Forged brass valve, floating stainless steel needle, replaceable stainless steel seat
$399.00USD / Each
ICI Quick Ship Laboratory Needle Valve, Four Turrets, Vacuum EW-78985-10 Mfr # CPL2880-134WSA-V
COMPARE
Laboratory Needle Valve, Four Turrets, Vacuum Four 90° Turrets Forged brass valve, floating stainless steel needle, replaceable stainless steel seat
$722.00USD / Each
Showing 1 - 20 of 32