ICI Quick Ship Work Surfaces
Work surfaces for a quick delivery furniture program that builds complete labs
  • An extensive inventory of high quality furnishings, chemical fume hoods, and components
  • Clear and easy project communication
  • Design, project management, and factory trained installation
Custom manufactured work surfaces can be ordered in different depths, with special edge details, with extra high backsplashes, or a variety of cutouts. Choose from phenolic or epoxy material to meet your application needs.
Phenolic work surfaces have good chemical resistance to a wide range of acids, alkalies, and solvents. The 1” thick cellulose fiber reinforced phenolic resin core has a polyurethane copolymer surface. The appearance is black or grey with a smooth non-glare surface. This surface material is best whenever a high degree of chemical resistance is required.
Epoxy work surfaces also have a good chemical resistance to a wide range of acids, alkalies, and solvents. However, they also excel in resistance to heat and sturdiness. The 1” thick molded epoxy resin appears black with a smooth, non-porous surface. This surface material is best whenever resistance to heat is required.
$39.55 - $2,192.00USD / Each
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
ICI Quick Ship Custom Manufactured Black Epoxy Work Surface EW-78984-50 Mfr # CPCMWSSF-E
COMPARE
Custom Manufactured Black Epoxy Work Surface
$112.50USD / Each
ICI Quick Ship Custom Manufactured Black Epoxy Work Surface Sink Cutout EW-78984-51 Mfr # CPCMWSSC-E
COMPARE
Custom Manufactured Black Epoxy Work Surface Sink Cutout
$116.30USD / Each
ICI Quick Ship Custom Manufactured Black Epoxy Work Surface Fixture/Boring Cutout EW-78984-52 Mfr # CPCMWSB-E
COMPARE
Custom Manufactured Black Epoxy Work Surface Fixture/Boring Cutout
$39.55USD / Each
ICI Quick Ship Custom Manufactured Black Phenolic Work Surface EW-78984-53 Mfr # CPCMWSSF-P
COMPARE
Custom Manufactured Black Phenolic Work Surface
$53.95USD / Each
ICI Quick Ship Custom Manufactured Black Phenolic Work Surface Sink Cutout EW-78984-54 Mfr # CPCMWSSC-P
COMPARE
Custom Manufactured Black Phenolic Work Surface Sink Cutout
$116.30USD / Each
ICI Quick Ship Custom Manufactured Black Phenolic Work Surface Fixture/Boring Cutout EW-78984-55 Mfr # CPCMWSB-P
COMPARE
Custom Manufactured Black Phenolic Work Surface Fixture/Boring Cutout
$39.55USD / Each
ICI Quick Ship Black Epoxy Work Surface, 36" Wide, 30" D EW-78984-56 Mfr # CPLE3630-1
COMPARE
Black Epoxy Work Surface, 36" Wide, 30" D
$833.50USD / Each
ICI Quick Ship Black Epoxy Work Surface, 48" Wide, 30" D EW-78984-57 Mfr # CPLE4830-1
COMPARE
Black Epoxy Work Surface, 48" Wide, 30" D
$1,103.50USD / Each
ICI Quick Ship Black Epoxy Work Surface, 60" Wide, 30" D EW-78984-58 Mfr # CPLE6030-1
COMPARE
Black Epoxy Work Surface, 60" Wide, 30" D
$1,379.00USD / Each
ICI Quick Ship Black Epoxy Work Surface, 72" Wide, 30" D EW-78984-59 Mfr # CPLE7230-1
COMPARE
Black Epoxy Work Surface, 72" Wide, 30" D
$1,655.50USD / Each
ICI Quick Ship Black Epoxy Work Surface, 96" Wide, 30" D EW-78984-60 Mfr # CPLE9630-1
COMPARE
Black Epoxy Work Surface, 96" Wide, 30" D
$2,192.00USD / Each
ICI Quick Ship Black Phenolic Work Surface, 36" Wide, 30" D EW-78984-61 Mfr # CPLP3630-1
COMPARE
Black Phenolic Work Surface, 36" Wide, 30" D
$398.40USD / Each
ICI Quick Ship Black Phenolic Work Surface, 48" Wide, 30" D EW-78984-62 Mfr # CPLP4830-1
COMPARE
Black Phenolic Work Surface, 48" Wide, 30" D
$542.25USD / Each
ICI Quick Ship Black Phenolic Work Surface, 60" Wide, 30" D EW-78984-63 Mfr # CPLP6030-1
COMPARE
Black Phenolic Work Surface, 60" Wide, 30" D
$678.25USD / Each
ICI Quick Ship Black Phenolic Work Surface, 72" Wide, 30" D EW-78984-64 Mfr # CPLP7230-1
COMPARE
Black Phenolic Work Surface, 72" Wide, 30" D
$814.00USD / Each
ICI Quick Ship Black Phenolic Work Surface, 96" Wide, 30" D EW-78984-65 Mfr # CPLP9630-1
COMPARE
Black Phenolic Work Surface, 96" Wide, 30" D
$1,153.50USD / Each
ICI Quick Ship Black Epoxy Work Surface Curb, 96" Wide, 4" High EW-78984-66 Mfr # CPLE9604-1
COMPARE
Black Epoxy Work Surface Curb, 96" Wide, 4" High
$299.80USD / Each
ICI Quick Ship Black Phenolic Work Surface Curb, 96" Wide, 4" High EW-78984-67 Mfr # CPLP9604-1
COMPARE
Black Phenolic Work Surface Curb, 96" Wide, 4" High
$144.30USD / Each
Showing 1- 18 of 18