This product is now discontinued, View Related Products

Idex® Debubblers for Liquid Chromatography

Eliminate interference from bubbles
  • Quickly, efficiently capture and remove dissolved gases
  • Reduce instrument downtime and improve performance
  • Integrate into any stand-alone degasser, degassing tray, or pump
  • Reduce partial reagent dispenses, creating fewer false positives
Integrating a debubbler into the fluid path creates stable instrument performance across systems using applications that require dissolved gas by capturing and removing bubbles. You can choose components to actively remove bubbles with or without removing dissolved gases in the system. Debubblers reduce downtime and waste due to bubble formation and improve performance across your system.

Models that include a degasser remove bubbles from the liquid stream by vacuum through a Poridex™ membrane. Degassing is ideal for preventing bubble formation from recurring downstream.
Call For Price

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Max Pressure (PSI) Max Temperature (° C) Membrane Material
Availability Pricing
Idex Debubbler, ETFE, No Degasser Tail; 2.5 mL EW-42712-33 Mfr # 9000-1540
COMPARE
30 40 Poridex
Call For Price
Idex 9000-1541 Debubbler, ETFE, No Degasser Tail; 5 mL EW-42712-34 Mfr # 9000-1541
COMPARE
30 40 Poridex
Call For Price
Idex 9000-1544 Debubbler, ETFE, with Degasser Tail; 2.5 mL EW-42712-35 Mfr # 9000-1544
COMPARE
30 40 Poridex
Call For Price
Idex 9000-1545 Debubbler, ETFE, with Degasser Tail; 5 mL EW-42712-36 Mfr # 9000-1545
COMPARE
30 40 Poridex
Call For Price
Idex 9000-1549 Transfer Line Debubbler, 4 mL, 43" L; 1/EA  EW-42712-37 Mfr # 9000-1549
COMPARE
30 40 Poridex
Call For Price
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat