This product is now discontinued, View Related Products
IDEX Plugs for 6-32 Ports
Use the plugs to close off unused ports in valves and multiport connectors. Select coned plugs for high-pressure applications; flat-bottom plugs for low-pressure applications.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Color Material Compatible With
Availability Pricing
Idex P-411 Port Plug, Microtight® Headless, Black PEEK, 6-32 Coned; 1/EA EW-02018-38 Mfr # P-411
COMPARE
Black PEEK/PCTFE 6-32 coned port
Call For Price
Idex P-559 Port Plug, Micro Headless, Natural PEEK, 6-32 Flat-Bottom; 1/EA EW-02018-47 Mfr # P-559
COMPARE
Black PEEK 6-32 flat-bottom port