IDEX® Precolumn Microfilters

Filter out particles before they get into analytical columns
  • Connect directly to columns with 10-32 threads
  • Made of PEEK® or stainless steel parts
  • Total theoretical void volume 0.5 µm
Connect these precolumn microfilters directly into your analytical columns or microbores and virtually eliminate harmful particles from clogging up your system. The PEEK tubing used in these assemblies has an internal diameter of 0.005" (125 µm), preventing the mobile phase from mixing with the sample. The total theoretical void volume is 0.5 µm, which includes the frit volume.

Microfilters are available to connect to MicroTight® tubing sleeves for 70 to 520 µm OD capillary tubing or select a version that connects directly to 1/16" OD tubing.
$133.75 - $164.75USD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Connections Pore Size (µm) Description
Availability Pricing
Idex M-500 Precolumn MicroFilter Assembly, Silica/SS, 0.5 µm, 1/4" OD Tubing, 10-32 Coned; 1/EA EW-02010-46 Mfr # M-500
COMPARE
MicroTight tubing sleeves 0.5 Precolumn MicroFilter Assembly, Silica/SS, 0.5 µm, 1/4\" OD Tubing, 10-32 Coned; 1/EA
$163.75USD / Each
Idex M-510 Precolumn MicroFilter Assembly, Silica/PEEK, 0.5 µm, 1/4" OD Tubing, 10-32 Coned; 1/EA EW-02010-48 Mfr # M-510
COMPARE
MicroTight tubing sleeves 0.5 Precolumn MicroFilter Assembly, Silica/PEEK, 0.5 µm, 1/4\" OD Tubing, 10-32 Coned; 1/EA
$157.50USD / Each
Idex M-550 Precolumn MicroFilter Assembly, PEEK/SS, 0.5 µm, 1/16" OD, 10-32 Coned; 1/EA EW-02010-49 Mfr # M-550
COMPARE
1/16\" OD tubing 0.5 Precolumn MicroFilter Assembly, PEEK/SS, 0.5 µm, 1/16\" OD, 10-32 Coned; 1/EA
$164.75USD / Each
Idex M-555-1 Precolumn MicroFilter, SS, 2 µm, 1/16" OD; 1/EA EW-02010-51 Mfr # M-555-1
COMPARE
1/16\" OD tubing 2 Precolumn MicroFilter, SS, 2 µm, 1/16\" OD; 1/EA
$133.75USD / Each
Idex M-560 Precolumn MicroFilter, PEEK, 0.5 µm, 1/16" OD Tubing, 10-32 Coned; 1/EA EW-02010-52 Mfr # M-560
COMPARE
1/16\" OD tubing 0.5 Precolumn MicroFilter, PEEK, 0.5 µm, 1/16\" OD Tubing, 10-32 Coned; 1/EA
$164.75USD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat