IDEX Ultra-Low Volume Back Pressure Regulators
Minimize swept volume in ultra-low volume applications
Ultra-low volume back pressure regulators have a swept volume of only 6 µL making it nearly impossible to detect them as part of your fluid pathway. Wetted flow path materials are PEEK, perfluoroelastomer, and ETFE. 1/4-28 flat-bottom fitting accepts 1/16" OD tubing.
$275.00 - $404.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Pressure Setting (PSI) Tubing OD (in) Working Pressure (PSI)
Availability Pricing
Idex M-412 Back Pressure Regulator, Ultra-Low Volume, 500 psi, 1 mL/min, 1/16" OD Tubing; 1/EA EW-02013-55 Mfr # M-412
COMPARE
500 0.0625 250 to 525 psi
$282.00USD / Each
Idex M-410 Back Pressure Regulator Assembly, Ultra-Low Volume, 100 psi, 1 mL/min, 1/16" OD Tubing; 1/EA EW-02017-06 Mfr # M-410
COMPARE
100 0.0625 40 to 150 psi
$275.00USD / Each
Idex M-420 Back Pressure Regulator Assembly, Ultra-Low Volume, 100 psi, 8 mL/min, 1/16" OD Tubing; 1/EA EW-02017-07 Mfr # M-420
COMPARE
100 0.0625 40 to 150 psi
$404.00USD / Each