IKA Incubating Shakers
Advanced temperature control and safety features enable unattended operation in a temperature-controlled environment
  • Antimicrobial coating reduces bacteria on control surface
  • Easy-to-read LED display of speed, temperature, and time
  • RS-232 and USB ports for PC interface
  • All functions can be controlled and documented using labworldsoft® software, (sold separately below)
Shakers features PID temperature control, to maintain temperature within ±0.1°C. Independently monitor temperature by connecting an additional PT 1000 temperature sensor to a junction box within the chamber. Electronic speed control can be set from 10 to 500 rpm with a 20-mm orbit. Shaker can be set for timed operation up to 999 hours or for continuous operation. Unit shuts off automatically when lid is lifted. Collecting tray with drain hose on rear of unit simplifies cleanup.
Platforms, attachments, and software are sold separately below.
$5,923.00 - $9,636.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Load (lbs) Power (VAC)
Availability Pricing
IKA 3510001 KS 4000i Control Incubating Shaker - 115VAC EW-51715-00 Mfr # 3510001
COMPARE
44 115
$8,776.00USD / Each
IKA 3940001 KS 3000 i Control Incubating Shaker, 115 VAC EW-51715-01 Mfr # 3940001
COMPARE
16.5 115
$5,923.00USD / Each
IKA 3940101 KS3000ic Incubating Shaker with Built-in Cooling Coil, 115VAC EW-51715-02 Mfr # 3940101
COMPARE
16.5 115
$7,404.00USD / Each
IKA 3510000 KS 4000i Control Incubating Shaker - 230VAC 50/60 Hz EW-51715-05 Mfr # 3510000
COMPARE
44 230
$8,776.00USD / Each
IKA 3510101 KS 4000ic Control Refrigerated Incubating Shaker - 115VAC EW-51715-10 Mfr # 3510101
COMPARE
44 115
$9,636.00USD / Each
IKA 3510100 KS 4000ic Control Refrigerated Incubating Shaker - 230VAC EW-51715-15 Mfr # 3510100
COMPARE
44 230
$9,636.00USD / Each