IKA Large Capacity Digital Stirrers
Digital control for your large, heavy-duty mixing
  • Effectively stirs up to 50 liters
Powerful magnetic stirrers feature a flat, sturdy stainless steel casing—ideal for your heavy-duty mixing needs. Digital LED display allows for exact control. Unit features a timer for up to 56 minutes or set for continuous run.
$5,962.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Top Plate Length (in)
Availability Pricing
IKA 2621801 19" x 19" Large Capacity Digital Stirrer, 115V EW-04671-82 Mfr # 25002981
COMPARE
115 19
$5,962.00USD / Each