IKA LR 1000 Reaction Stations
Optimize chemical reactions in your laboratory with these modular reaction systems
  • Digital display for monitoring speed and temperature
  • Included pH monitoring and weighing function (control model only)
  • RS-232 and USB connections for PC control (control model includes an option for Bluetooth)
Both systems are used to reproduce and enhance chemical reactions as well as to mix, disperse, and homogenize. They have a wide range of uses from general laboratory to pharmaceutical. A digital display allows the user to reach a set temperature and speed in a controlled manner. Ports for output let you control and monitor your reaction right from your PC.
Control models present the addition of pH and weight measurement. Bluetooth interface and separate wireless controller added to increase convenience of use.
$15,040.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Display Type Control Type Power (VAC)
Availability Pricing
IKA LR 1000 Basic Reaction Station; 115/230 VAC EW-28710-98 Mfr # 0008040101
COMPARE
Basic 115 / 230
$15,040.00USD / Each